Skip to Content
Merck
All Photos(1)

Documents

K0254

Sigma-Aldrich

Kanamycin solution from Streptomyces kanamyceticus

liquid, suitable for cell culture and plant cell culture, BioReagent

Synonym(s):

Kanamycin A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

product name

Kanamycin solution from Streptomyces kanamyceticus, 50 - 60 mg/mL in 0.9% NaCl, BioReagent, liquid, 0.1 μm filtered, suitable for cell culture

biological source

Streptomyces kanamyceticus

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

concentration

50 - 60 mg/mL in 0.9% NaCl

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17/h4-18,23-29H,1-3,19-22H2/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-/m1/s1

InChI key

SBUJHOSQTJFQJX-NOAMYHISSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kanamycin sulfate is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 2 mL/L.

Application

Kanamycin solution from Streptomyces kanamyceticus has been used:
  • to induce deafness in neonatal rats
  • as an endothelial growth medium component for culturing human umbilical vein endothelial cells (HUVECs)
  • as medium component for Mycobacteria Cultures and Top10 E coli cells

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance: Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Caution

Solutions are stable at 37°C for approximately 5 days. Aqueous stock solutions can be stored at 2-8°C for long term storage.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrocyte Hypertrophy and Microglia Activation in the Rat Auditory Midbrain Is Induced by Electrical Intracochlear Stimulation
Rosskothen-Kuhl N, et al.
Frontiers in Cellular Neuroscience, 12, 43-43 (2018)
Toll-like receptor 2 release by macrophages: an anti-inflammatory program induced by glucocorticoids and lipopolysaccharide
Hoppstadter J, et al.
Frontiers in immunology, 10, 1634-1634 (2019)
Generation of isogenic human iPS cell line precisely corrected by genome editing using the CRISPR/Cas9 system
Grobarczyk B, et al.
Stem Cell Reviews and Reports, 11(5), 774-787 (2015)
Indrikis A Krams et al.
The Journal of experimental biology, 220(Pt 22), 4204-4212 (2017-09-25)
Communities of symbiotic microorganisms that colonize the gastrointestinal tract play an important role in food digestion and protection against opportunistic microbes. Diet diversity increases the number of symbionts in the intestines, a benefit that is considered to impose no cost
Samuel S Porter et al.
Current protocols in microbiology, 57(1), e101-e101 (2020-05-08)
This protocol describes the production of human papillomavirus (HPV)-derived quasiviruses. Quasiviruses are infectious particles that are produced in 293TT packaging cells and contain a complete viral genome. We describe methods for infection of primary human keratinocytes with HPV quasiviruses, as

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service