GBL470635
Grace Bio-Labs ProPlate® microarray system, slide module only
well size 3, L × W × depth 20 mm × 20 mm × 7.6 mm, without clips
Synonym(s):
incubation chambers, microarray hybridization chambers, slide chambers
Select a Size
$198.00
Select a Size
About This Item
$198.00
Recommended Products
material
black silicone (on Delrin)
feature
without clips
packaging
pack of 10 ea
manufacturer/tradename
Grace Bio-Labs 470635
L × W × depth
20 mm × 20 mm × 7.6 mm
well size
3
Related Categories
General description
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
- Up to 64 leak-proof wells on any slide surface without the use of adhesive
- Microtiter plate well spacing facilitates use with multichannel pipettes
- 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)