Key Documents
AXYPCR0208C100
Corning® Axygen® PCR Tube Strips
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
UNSPSC Code:
24122002
NACRES:
NB.22
Pricing and availability is not currently available.
manufacturer/tradename
Corning PCR-0208-C-100
Related Categories
General description
  • Corning® Axygen® PCR tube strips are made of 99.9% pure virgin polypropylene.
  • Designed to fit a wide variety of thermal cyclers.
  • Thin, consistent, and uniform wall thickness facilitate accurate thermal transfer and optimal amplification.
  • An even insertion cap prevents evaporation and cross-contamination of the samples.
  • Tubes are free of RNase-/DNase and nonpyrogenic.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.