Key Documents
Z280372
Sigma-Aldrich
PVC laboratory tubing
I.D. × O.D. 1/4 in. × 3/8 in.
Synonym(s):
Saint Gobain Versilon ABH02017
Select a Size
100 FT
$86.30
$86.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 FT
$86.30
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
$86.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 100 feet
I.D. × O.D.
1/4 in. × 3/8 in.
wall size
1/16 in.
General description
General-purpose polyvinyl chloride laboratory tubing. Clear, flexible tubing ideal for drains and rinse tubes. Max. temp.: 165°F (73.4°C).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Is this tubing autoclavable?
    1 answer
    1. The material can withstand a standard autoclave cycle (steam 30 minutes at 15 psi, 250° F (121° C)), but for repeated sterilization, this product would not be suitable.
      Helpful?
  2. Could you provide information about the maximum psi of PVC laboratory tubing Z280372-1PAK?
    1 answer
    1. The maximum operating pressure is 30 pounds per square inch at 73 degrees Fahrenheit.
      Helpful?