Skip to Content
Merck
All Photos(2)

Key Documents

G9287

Sigma-Aldrich

Globotriose

Synonym(s):

α-D-Gal-(1→4)-β-D-Gal-(1→4)-D-Glc, 4-O-(4-O-α-D-Galactopyranosyl-β-D-galactopyranosyl)-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥95% (HPLC)

Quality Level

form

powder

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O[C@@H]2[C@H](O)[C@@H](O)[C@@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7+,8+,9-,10-,11-,12-,13-,14-,15+,16?,17-,18+/m1/s1

InChI key

FYGDTMLNYKFZSV-ANKSBSNASA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H C Hansen et al.
Carbohydrate research, 322(3-4), 166-180 (2000-01-19)
Six aminodeoxy 2-(trimethylsilyl)ethyl (Me3SiEt) glycoside analogues of galabiose (4'- and 6'-aminodeoxy) and globotriose (6"-, 4"-, 2"-, and 6'-aminodeoxy) were synthesized by glycosylation of protected Me3SiEt galactoside and lactoside acceptors with azido-substituted monosaccharide donors, followed by reduction of the azido groups
Hirofumi Dohi et al.
Carbohydrate research, 337(11), 983-989 (2002-06-01)
New thioglycosyl donors (o-methoxycarbonylphenyl 2,3,4,6-tetra-O-benzyl-1-thio-beta-D-galactopyranoside and its 6-O-acetyl analogue) were designed and used for the synthesis of glycoconjugate polymers carrying Gb(3) [Gal(alpha1-->4)Gal(beta1-->4)Glc] and isoGb(3) [Gal(alpha1-->3)Gal(beta1-->4)Glc] clusters as side chains. These donors scarcely evolved the unpleasant odor of thiophenols and showed
James L Leach et al.
Antimicrobial agents and chemotherapy, 49(9), 3842-3846 (2005-08-30)
Epithelial cells lining the urinary tract are rich in globo series glycolipids, structurally defined by a Galalpha1,4Gal motif in the oligosaccharide moiety of this glycolipid family. This Galalpha1,4Gal motif is the attachment target for the P-fimbrial adhesin of uropathogenic Escherichia
Hussein Masoud et al.
Biochemistry, 42(15), 4463-4475 (2003-04-16)
Lipopolysaccharide (LPS) is a major virulence determinant of the human bacterial pathogen Haemophilus influenzae. Structural elucidation of the LPS from H. influenzae type b strain RM7004 was achieved by using electrospray ionization mass spectrometry (ESI-MS) and high-field NMR techniques on
Xuebing Li et al.
Journal of medicinal chemistry, 55(6), 2702-2710 (2012-03-01)
Shiga toxin (Stx)-producing Escherichia coli (STEC) causes diarrhea and colitis in humans that can develop into a life-threatening hemolytic uremic syndrome (HUS). Developing efficient means of controlling STEC diseases, for which no drugs or vaccines are currently available, remains a

Articles

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service