Skip to Content
Merck
All Photos(1)

Key Documents

G2921

Sigma-Aldrich

Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides

BioUltra, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥95% (SDS-PAGE), ≥550 units/mg protein (biuret)

Synonym(s):

G-6-P-DH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

product line

BioUltra

Assay

≥95% (SDS-PAGE)

form

ammonium sulfate suspension

specific activity

≥550 units/mg protein (biuret)

foreign activity

Creatine Phosphokinase, Phosphoglucose isomerase, lactic dehydrogenase, glutathione reductase, NADH oxidase, NADPH oxidase, phosphoglucomutase, 6-phosphogluconic dehydrogenase, hexokinase ≤0.001%
Myokinase ≤0.005%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Glucose-6-phosphate dehydrogenase (G6PD) is an X-linked essential enzyme that protects cells from oxidative stress, particularly red blood cells.

Application

Glucose-6-phosphate dehydrogenase was used to study atypical glycolysis in Clostridium thermocellum.

Biochem/physiol Actions

Glucose-6-phosphate dehydrogenase (G6PD) catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway.
Glucose-6-phosphate dehydrogenase catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway.

Unit Definition

One unit will oxidize 1.0 μmole of D-glucose-6-phosphate to 6-phospho-D-gluconate per minute in the presence of NAD at pH 7.8 at 30°C

Physical form

Supplied in 3.2M ammonium sulfate containing 50mM Tris and 1mM magnesium chloride, pH 7.5

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jilai Zhou et al.
Applied and environmental microbiology, 79(9), 3000-3008 (2013-02-26)
Cofactor specificities of glycolytic enzymes in Clostridium thermocellum were studied with cellobiose-grown cells from batch cultures. Intracellular glucose was phosphorylated by glucokinase using GTP rather than ATP. Although phosphofructokinase typically uses ATP as a phosphoryl donor, we found only pyrophosphate
Signs of endotracheal intubation in a field setting.
R D Stewart et al.
Annals of emergency medicine, 14(3), 276-277 (1985-03-01)
L Zuluaga et al.
Experimental parasitology, 133(1), 114-120 (2012-11-28)
Dehydroepiandrosterone (DHEA) inhibits glucose 6-phosphate dehydrogenase (G6PDH) of different species and may potentially decrease intracellular glutathione. Therefore, it can have and enhance anti-parasitic action against Plasmodium spp. We evaluated the antiplasmodial activity and the interaction of DHEA with several antimalarial
Clara Slade Oliveira et al.
Reproduction (Cambridge, England), 145(1), 9-17 (2012-10-30)
During initial development, both X chromosomes are active in females, and one of them must be silenced at the appropriate time in order to dosage compensate their gene expression levels to male counterparts. Silencing involves epigenetic mechanisms, including histone deacetylation.
Peter A Hecker et al.
Circulation. Heart failure, 6(1), 118-126 (2012-11-22)
Glucose 6-phosphate dehydrogenase (G6PD) is the most common deficient enzyme in the world. In failing hearts, G6PD is upregulated and generates reduced nicotinamide adenine dinucleotide phosphate (NADPH) that is used by the glutathione pathway to remove reactive oxygen species but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service