Skip to Content
Merck
All Photos(1)

Documents

F8646

Sigma-Aldrich

Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:320

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody is specific for the Fc fragment of mouse IgG. No cross reactivity is observed with the Fab fragment of mouse IgG. Purified antibody is conjugated to fluorescein isothiocyanate (FITC) in an alkaline reaction and then purified to remove unbound FITC.

Specificity

Binds mouse IgG; does not bind other mouse Igs.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody may be used for immunohistochemistry at a working dilution of 1:320 using formalin-fixed, paraffin-embedded sections of human tonsil. Antibody dilution of 1:200 was used to label sections of human brainstems and cervical spinal cords with primary antibody against human Bcl-2 protein. The antibody was also used to label erythrocytes for photon reassignment microscopy and for FACS using melanoma cells.

Other Notes

Antibody adsorbed with bovine, equine, and human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with bovine, equine, or human samples.
Useful when trying to avoid background staining due to the presence of Fc receptors.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Muczynski et al.
Journal of immunology (Baltimore, Md. : 1950), 160(7), 3207-3216 (1998-04-08)
We compared HLA class II expression in a human melanoma line (a nonprofessional APC), induced by IFN-gamma or by stable transfection with CIITA, with constitutive class II expression in an EBV-transformed B lymphoblastoid cell line (a professional APC) from the
J Gysin et al.
Infection and immunity, 67(12), 6596-6602 (1999-11-24)
We performed ex vivo experiments with Plasmodium falciparum-infected human placentas from primi- and multigravida women from Cameroon. All women, independent of their gravida status, had anti-chondroitin sulfate A (CSA) adhesion antibodies which cross-reacted with heterologous strains, such as FCR3 and
M Coulpier et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 16(19), 5897-5904 (1996-10-01)
The molecular events leading to motoneuronal death are still poorly understood. In mammals, the bcl-2 proto-oncogene, which encodes a membrane-associated protein, has been shown to suppress both developmental motoneuronal death and experimental axotomy-induced motoneuronal death. We assessed a potential protective
Gabrielle Dias Salton et al.
Cancer biology & therapy, 18(8), 560-570 (2017-07-12)
Eukaryote initiation factor 2 subunit β (eIF2β) plays a crucial role in regulation protein synthesis, which mediates the interaction of eIF2 with mRNA. eIF2β contains evolutionarily conserved polylysine stretches in amino-terminal region and a zinc finger motif in the carboxy-terminus.
Daniel Sehic et al.
American journal of clinical pathology, 141(6), 828-833 (2014-05-20)
Successful further treatment of Wilms tumors (WTs) after preoperative chemotherapy and surgery depends on correct histopathologic risk stratification, including quantification of remaining blastemal elements. In the present study, we assessed the usefulness of protein markers for the detection of WT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service