F7177
Nunc® EasYFlasks™, Nunclon® Δ Surface
Nunclon® Δ surface treatment, 25 cm2 angled neck, vent/close cap, 200/cs
Synonym(s):
cell culture flask, tissue culture flask, tissue culture treated flask
Select a Size
$599.00
Estimated to ship onApril 11, 2025
Select a Size
About This Item
$599.00
Estimated to ship onApril 11, 2025
Recommended Products
material
canted neck transparent polystyrene flask
screw top blue high-density polyethylene cap (vented)
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 200 (20 packs x 10 flasks)
manufacturer/tradename
Nunc 156340
surface area
25 cm2
working volume
7 mL
fitting
25 mm (1 in.) cap
binding type
Nunclon Delta surface
General description
- Nunclon® Δ certified: a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
- Ergonomic design with a 1/3 turn to open, close or vent
- Low profile to maximize incubator space
- Volume graduations printed and molded on both sides
- Slightly angled side walls for complete viewing under a microscope
- Sterilized by gamma irradiation and certified to be nonpyrogenic
- Nunclon Delta Certification, including testing on four different cell types, provided in each case
- Vent/close caps feature a “Y” mark that allows visual verification of vent position.
- Vent position easily distinguished even when flasks are stacked in incubators.
- Filter membrane provides effective barrier against micro-organisms.
- Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation.
- Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)