Skip to Content
Merck
All Photos(1)

Key Documents

F5884

Sigma-Aldrich

α-L-Fucosidase from bovine kidney

ammonium sulfate suspension, ≥2.0 units/mg protein (biuret)

Synonym(s):

α-L-Fucoside fucohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

bovine kidney

form

ammonium sulfate suspension

specific activity

≥2.0 units/mg protein (biuret)

mol wt

210-220 kDa (Subunits: 55-60 kDa)

foreign activity

α-mannosidase and β-galactosidase ≤0.1%
β-N-acetylglucosaminidase ≤0.2%

shipped in

wet ice

storage temp.

2-8°C

General description

α-L-fucosidases is a liposomal enzyme, which belongs to glycosyl hydrolases family.

Application

α-L-fucosidase from bovine kidney has been used:
  • to study their ability to alter primary cell transduction
  • to examine the role of fucose residues of cell surface glycans in adhesion
  • to investigate its role during fertilization

Biochem/physiol Actions

α-L-fucosidases degrades fucose-containing fucoglycoconjugates and breaks fucosidic bonds in oligosaccharides and glycoconjugates. It acts as a tumor marker for hepatic and colorectal cancer. α-L-fucosidase exhibits transglycosylation property.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl α-L-fucoside to p-nitrophenol and L-fucose per min at pH 5.5 at 25°C.

Physical form

Suspension in 3.2 M (NH4)2SO4, 10 mM NaH2PO4 10 mM citrate, pH 6.0

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel alpha-l-fucosidases from a soil metagenome for production of fucosylated human milk oligosaccharides
Lezyk M, et al.
PLoS ONE, 11(1), e0147438-e0147438 (2016)
Human alpha-L-fucosidase-1 attenuates the invasive properties of thyroid cancer
Vecchio G, et al.
Testing, 8(16), 27075-27075 (2017)
Directed evolution of the alpha-L-fucosidase from Thermotoga maritima into an alpha-L-transfucosidase
Osanjo G, et al.
Biochemistry, 46(4), 1022-1033 (2007)
alpha-l-Fucosidase enhances capacitation-associated events in porcine spermatozoa
Romero-Aguirregomezcorta J, et al.
The Veterinary Journal, 203(1), 109-114 (2015)
Alline R Pacheco et al.
Nature, 492(7427), 113-117 (2012-11-20)
The mammalian gastrointestinal tract provides a complex and competitive environment for the microbiota. Successful colonization by pathogens requires scavenging nutrients, sensing chemical signals, competing with the resident bacteria and precisely regulating the expression of virulence genes. The gastrointestinal pathogen enterohaemorrhagic

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service