Skip to Content
Merck
All Photos(1)

Key Documents

F6272

Sigma-Aldrich

α-1,6-Fucosidase solution from Elizabethkingia miricola

recombinant, expressed in E. coli, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.04 UNIT
$809.00

$809.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
0.04 UNIT
$809.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$809.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥1.8 units/mg protein

UniProt accession no.

foreign activity

β-galactosidase, α-mannosidase, β-hexosaminidase, neuraminidase and proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Biochem/physiol Actions

For reactions with complex carbohydrates, the substrate must be labeled with aminonaphthalenetrisulfonic acid (ANTS).

Unit Definition

One unit will release 1.0 μmole of methylumbelliferone from 4-methylumbelliferyl α-L-fucoside per min at pH 5.0 at 37 °C

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 25 mM NaCl

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service