Skip to Content
Merck
All Photos(1)

Key Documents

F3883

Supelco

Flurazepam dihydrochloride

Synonym(s):

7-Chloro-1-(diethylamino)ethyl-5-(2-fluorophenyl)-3H-1,4-benzodiazepin-2(1H)-one dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFN3O · 2HCl
CAS Number:
Molecular Weight:
460.80
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

2-hydroxypropyl-β-cyclodextrin: 6 mg/mL
H2O: soluble

format

neat

SMILES string

Cl[H].Cl[H].CCN(CC)CCN1C(=O)CN=C(c2ccccc2F)c3cc(Cl)ccc13

InChI

1S/C21H23ClFN3O.2ClH/c1-3-25(4-2)11-12-26-19-10-9-15(22)13-17(19)21(24-14-20(26)27)16-7-5-6-8-18(16)23;;/h5-10,13H,3-4,11-12,14H2,1-2H3;2*1H

InChI key

ZIIJJOPLRSCQNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anxiolytic; anticonvulsant; GABAA receptor benzodiazepine modulatory site ligand.
Benzodiazepine anxiolytic; anticonvulsant; ligand for the GABAA receptor benzodiazepine modulatory site.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E I Tietz et al.
Neuroscience, 91(1), 327-341 (1999-05-21)
The effect of prolonged benzodiazepine administration on GABA(A) receptor subunit (alpha1-6, beta1-3, gamma2) messenger RNAs was investigated in the rat hippocampus and cortex, among other brain areas. Rats were orally administered flurazepam for one week, a protocol which results in
P Poisbeau et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(10), 3467-3475 (1997-05-15)
Whole-cell patch-clamp recordings were made from CA1 pyramidal and dentate gyrus granule cells (GCs) in hippocampal slices to assess the effects of withdrawal from chronic flurazepam (FRZ) treatment on the function of synaptic GABAA receptors. In slices from control rats
Marie-Pierre Sylvestre et al.
Statistics in medicine, 28(27), 3437-3453 (2009-08-27)
Many epidemiological studies assess the effects of time-dependent exposures, where both the exposure status and its intensity vary over time. One example that attracts public attention concerns pharmacoepidemiological studies of the adverse effects of medications. The analysis of such studies
Guofu Shen et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 35(9), 1897-1909 (2010-05-07)
Benzodiazepine withdrawal anxiety is associated with potentiation of alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionate receptor (AMPAR) currents in hippocampal CA1 pyramidal neurons attributable to increased synaptic incorporation of GluA1-containing AMPARs. The contribution of calcium/calmodulin-dependent protein kinase II (CaMKII) to enhanced glutamatergic synaptic strength during withdrawal
Stephen I Deutsch et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 19(6), 398-401 (2009-02-05)
Stress induces changes in the endogenous tone of both GABA and NMDA receptor-mediated neurotransmission in the intact mouse. Because changes are observed 24 h after stress, epigenetically-regulated alterations in gene expression may mediate these effects. In earlier work, sodium butyrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service