Recommended Products
grade
analytical standard
packaging
pkg of 1 ea
manufacturer/tradename
Air Liquide America Specialty Gases LLC 16-05-129SP
technique(s)
gas chromatography (GC): suitable
General description
For use with Air Liquide′s Scotty Gas calibration standards supplied in the 14L and 48L cylinders. Additional septa may ordered using p/n 608010.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service