Skip to Content
Merck
All Photos(1)

Documents

EP5943000548

Eppendorf® Centrifuge 5910 Ri G

refrigerated, with S-4x750 rotor, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug

Synonym(s):

Eppendorf® Centrifuge 5910 Ri G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CE compliant
CSA approved
refrigerated

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5943000548

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Centrifuge 5910 Ri G
  • refrigerated
  • with touchscreen interface
  • with rotor S-4x750 incl. round buckets and adapters for 5/15/50 ml/plates
  • 120V, US

Application

Centrifugation

Features and Benefits

Super versatile multipurpose centrifuge with high capacity for broad range of consumables, refrigerated for temperature sensitive samples

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service