Skip to Content
Merck
All Photos(1)

Key Documents

EP0030723112

Eppendorf® Cell Culture Plates

size 48 wells, flat bottom clear wells, pkg of 60 plates (individually wrapped), sterile, lid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

flat bottom clear wells
polystyrene

sterility

sterile

feature

lid

packaging

pkg of 60 plates (individually wrapped)

manufacturer/tradename

Eppendorf® 0030723112

technique(s)

PCR: suitable

W × D × H

127.8 mm × 85.5 mm × 20.0 mm

size

48 wells

surface area

8.56 cm2

well diam.

10.4 mm

well volume

1.5 mL

working volume

0.3-0.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf Cell Imaging Dishes ensure excellent results in high resolution microscopy of living and fixed cells. The bottom of the 35 mm dishes is made from cover glass.
The central 18 mm cavity is lower than the level of the surrounding polystyrene dish, which enables the concentration of the cells during seeding on the glass surface and helps to reduce costs of antibodies and dyes. Plate handling and reference are easy, safe, and convenient due to a polygonal gripping zone and directional orientation marking.

Rigorous quality assurance for Eppendorf Cell Imaging Consumables includes

  • Proof on non-cytotoxicity according to ISO 10993 specifications
  • Lot-certified sterility according to ISO 11137 and 11135
  • Sterility assurance level (SAL) of 10-6
  • Lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase
  • Testing for cell attachment and cell growth with an anchorage-dependent cell line
  • Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic

Features and Benefits

Easy and fast well identification by contrast rich individual well ID and OptiTrack alphanumeric labeling

Moat surrounding the outer wells to prevent the “edge effect” when filled with liquid

Chimney-well design enables filling of the inter-well spaces of the plate to level out well-to-well temperature shifts outside the incubator

Robust stacking performance by pronounced rims on plate lid and excellent fitting of lid and base when used in stacks

Lid taps under the plate for minimized surface contact and therefore reduced risk of contamination

Easy differentiation of lid and plate bottom due to a pronounced corrugation and wider sized base

Pronounced ventilation gaps for optimized gas and temperature transfer when incubating in stacks

Optimized microscopical performance due to enhanced planarity, reduction of meniscus and clarity of the material

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service