Skip to Content
Merck
All Photos(1)

Key Documents

EP0030505204

Eppendorf® Deepwell plates, standard, 96 wells

white plate, conical bottom colorless wells, capacity 1000 μL, pkg of 80 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom colorless wells
polypropylene
white plate
capacity:
1000 μL
size:
96 wells
working volume:
1 mL
sterility:
sterile; autoclaved
Pricing and availability is not currently available.

material

conical bottom colorless wells
polypropylene
white plate

sterility

sterile; autoclaved

packaging

pkg of 80 ea

manufacturer/tradename

Eppendorf® 0030505204

availability

available only in (India)

technique(s)

PCR: suitable

capacity

1000 μL

size

96 wells

working volume

1 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® Deepwell plates, standard, 96 wells, white plate of 1000 μL capacity are available as PCR clean and used for sample storage/preparation.
  • They are manufactured from high-quality polypropylene which guarantees high resistance to chemicals, mechanical stress and temperature extremes.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling.
  • RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties.
  • Minimal remaining volume and high uniformity from well to well.
  • Raised well rims and smooth surface for reliable closing, even with heat sealing.
  • Easy and reliable stacking.
  • g-Safe®: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality.
  • High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes.
  • Manufactured without slip agents, plasticizers, and biocides-Materials which could have a negative effect on bioassays.
  • Autoclavable (121 °C, 20 min).

For greater reliability, stability and efficiency, choose Eppendorf Deepwell 96 well Plates. Available with white, yellow, green or blue frames, these deepwell plates offer high performance for all applications - both manual and automated. Thanks to OptiTrack matrix, a high-contrast alphanumeric label laser-engraved into the frame, you can find your sample up to 30% faster and reduce pipetting errors. For every application, from sample storage at 86 C to DNA denaturation at 100 C, rely on Eppendorf Deepwell Plates and set new standards in your laboratory.

Application

  • Sample storage and preparation Bacteria and yeast cultivation
  • High-throughput DNA and RNA isolation
  • Storage of cDNA or genomic libraries
  • Storage of siRNA or oligonucleotide libraries
  • Protein analysis Storage of valuable protein and DNA samples in Eppendorf LoBind Plates
  • High-throughput plasmid purification Creation of dilution series

Features and Benefits

  • OptiTrack matrix: up to 30 % faster sample detection and fewer pipetting errors thanks to high-contrast alphanumeric labeling
  • RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • g-Safe : exceptional centrifugation resistance up to 6,000 × g for faster protocols and improved sample quality
  • Raised corrugated edges and smooth surface for reliable sealing, even during heat sealing
  • Easily and reliably stackable
  • Minimal residual volume and high homogeneity from well to well
  • High-quality polypropylene for high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without mold release agents, plasticizers and biocides – substances that can negatively affect bioassays
  • Autoclavable (121 C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service