Skip to Content
Merck
All Photos(1)

Key Documents

EHU081441

Sigma-Aldrich

MISSION® esiRNA

targeting human MAPK14

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TATTTGGGCCAAGGTGTTTCCATTTCTCAATCAGTGCAGTGATACATGTACTCCAGAGGGACAGGGTGGACCCCCTGAGTCAACTGGAGCAAGAAGGAAGGAGGCAGACTGATGGCGATTCCCTCTCACCCGGGACTCTCCCCCTTTCAAGGAAAGTGAACCTTTAAAGTAAAGGCCTCATCTCCTTTATTGCAGTTCAAATCCTCACCATCCACAGCAAGATGAATTTTATCAGCCATGTTTGGTTGTAAATGCTCGTGTGATTTCCTACAGAAATACTGCTCTGAATATTTTGTAATAAAGGTCTTTGCACATGTGACCACATACGTGTTAGGAGGCTGCATGCTCTGGAAGCCTGGACTCTAAGCTGGAGCTCTTGGAAGAGCTCTTCGGTTTCTGAGCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan-Juan Song et al.
Pathology, research and practice, 213(10), 1282-1288 (2017-09-17)
This study was to identify the biomarkers for the malignancy and poor prognosis in patients with ovarian cancer. The protein expression of p38MAPK family isoform p38α (p38α) and activating transcription factor 2 (ATF2) was measured in 120 ovarian serous adenocarcinomas
Yumei Yan et al.
Scientific reports, 6, 37052-37052 (2016-11-16)
Hepatocellular carcinoma (HCC) is refractory to chemotherapies, necessitating novel effective agents. The lysosome inhibitor Bafilomycin A1 (BafA1) at high concentrations displays cytotoxicity in a variety of cancers. Here we show that BafA1 at nanomolar concentrations suppresses HCC cell growth in
Yu Wang et al.
Oncology reports, 39(1), 61-70 (2017-11-09)
Photodynamic therapy (PDT) is considered to be an advancing antitumor technology. PDT using hydrophilic/lipophilic tetra‑α-(4-carboxyphenoxy) phthalocyanine zinc (TαPcZn-PDT) has exhibited antitumor activity in Bel-7402 hepatocellular cancer cells. However, the manner in which p38 MAPK and caspase-9 are involved in the regulation
Zhe Zhang et al.
Virology, 508, 150-158 (2017-05-26)
Enterovirus71 (EV71) is the major causative agent of hand, foot and mouth disease, which threatens the health of infants and young children. The expression of inflammatory cytokines induced by this viral infection aggravate the illness. Here, we describe the anti-EV71
Y Kim et al.
The British journal of dermatology, 179(3), 689-701 (2018-02-28)
Adiponectin is an adipocyte-derived cytokine that circulates as a full-length protein and a fragment containing the globular domain of adiponectin (gAd). A recent study has reported the antimelanogenic effects of full-length adiponectin. To examine the involvement of gAd in melanogenesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service