Skip to Content
Merck
All Photos(1)

Key Documents

EHU069421

Sigma-Aldrich

MISSION® esiRNA

targeting human BAX

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGCTTCAGGGTTTCATCCAGGATCGAGCAGGGCGAATGGGGGGGGAGGCACCCGAGCTGGCCCTGGACCCGGTGCCTCAGGATGCGTCCACCAAGAAGCTGAGCGAGTGTCTCAAGCGCATCGGGGACGAACTGGACAGTAACATGGAGCTGCAGAGGATGATTGCCGCCGTGGACACAGACTCCCCCCGAGAGGTCTTTTTCCGAGTGGCAGCTGACATGTTTTCTGACGGCAACTTCAACTGGGGCCGGGTTGTCGCCCTTTTCTACTTTGCCAGCAAACTGGTGCTCAAGGCTGGCGTGAAATGGCGTGATCTGGGCTCACTGCAACCTCTGCCTCCTGGGTTCAAGCGATTCACCTGCCTCAGCATCCCAAGGAGCTGGGATTACAGGCCCTGTGCACCAAGGTGCCGGAACTGATCAGAACCATCATGGGCTGGACATTGGACTTCCTCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... BAX(581) , BAX(581)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-han Zhang et al.
Apoptosis : an international journal on programmed cell death, 21(4), 473-488 (2016-01-16)
Epirubicin (EPI) is widely used for triple negative breast cancer (TNBC), but a substantial number of patients develop EPI resistance that is associated with poor outcome. The underlying mechanism for EPI resistance remains poorly understood. We have developed and characterized
Ilaria Penna et al.
Oncotarget, 7(4), 3947-3965 (2015-12-19)
Intrinsic cross-resistance to inhibition of different signaling pathways may hamper development of combinatorial treatments in melanoma, but the relative frequency of this phenotype and the strategies to overcome this hurdle remain poorly understood. Among 49 BRAF-mutant melanoma cell lines from
Chia-Hui Huang et al.
Toxicology and applied pharmacology, 334, 35-46 (2017-09-05)
Quinacrine, which is clinically used as an antimalarial drug, has anti-cancer activity. However, mechanism underlying its cytotoxic effect remains to be completely elucidated. In the present study, we investigated the cytotoxic effect of quinacrine on human leukemia U937 cells. Quinacrine-induced
Bhaskar Saha et al.
Oncotarget, 8(43), 73905-73924 (2017-11-02)
In view of the inadequacy of neuroblastoma treatment, five hydroxystilbenes and resveratrol (Resv) were screened for their cytotoxic property against human neuroblastoma cell lines. The mechanism of cytotoxic action of the most potent compound
Desheng Zhong et al.
Journal of cellular biochemistry, 115(9), 1624-1635 (2014-05-03)
Pan-Bcl-2 family inhibitor obatoclax has been demonstrated to be effective against various cancers, of which the mechanism of action is not fully understood. In this study, we demonstrate that obatoclax suppressed esophageal cancer cell viability with concomitant G1/G0-phase cell cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service