Skip to Content
Merck
All Photos(1)

Key Documents

EHU074041

Sigma-Aldrich

MISSION® esiRNA

targeting human PRKAA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCTCAGAAGAGGAAGTTCTCAGCTGTCTTTACAACAGAAATCACCAGGATCCTTTGGCAGTTGCCTACCATCTCATAATAGATAACAGGAGAATAATGAATGAAGCCAAAGATTTCTATTTGGCGACAAGCCCACCTGATTCTTTTCTTGATGATCATCACCTGACTCGGCCCCATCCTGAAAGAGTACCATTCTTGGTTGCTGAAACACCAAGGGCACGCCATACCCTTGATGAATTAAATCCACAGAAATCCAAACACCAAGGTGTAAGGAAAGCAAAATGGCATTTAGGAATTAGAAGTCAAAGTCGACCAAATGATATTATGGCAGAAGTATGTAGAGCAATCAAACAATTGGATTATGAATGGAAGGTTGTAAACCCATATTATTTGCGTGTACGAAGGAAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuiping Dai et al.
Oncotarget, 8(56), 95810-95823 (2017-12-10)
LB-100 is a novel PP2A inhibitor. Its activity in human colorectal cancer (CRC) cells was tested. The
Takuma Hashimoto et al.
Biochemical and biophysical research communications, 505(1), 13-19 (2018-09-19)
Solid tumors often contain hypoxic regions because an abnormal and inefficient tumor vasculature is unable to supply sufficient oxygen. Tissue hypoxia is generally defined as a low oxygen concentration of less than 2%. It is well known that tumor cells
Vanessa Zurli et al.
Science signaling, 13(631) (2020-05-14)
Understanding the costimulatory signaling that enhances the activity of cytotoxic T cells (CTLs) could identify potential targets for immunotherapy. Here, we report that CD2 costimulation plays a critical role in target cell killing by freshly isolated human CD8+ T cells
You-Li Yao et al.
Toxicology letters, 281, 127-138 (2017-10-02)
The aim of this study was to investigate the effects of acanthoic acid (AA) on the regulation of inflammatory response, lipid accumulation, and fibrosis via AMPK- IRAK4 signaling against chronic alcohol consumption in mice. Ethanol-induced liver injury was induced in
Ying-Nan Wang et al.
Oncogene, 39(3), 637-650 (2019-09-19)
Patients with stage II or III colorectal cancer (CRC) exhibit various clinical outcomes after radical treatments. The 5-year survival rate was between 50 and 87%. However, the underlying mechanisms of the variation remain unclear. Here we show that AMPKα1 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service