Skip to Content
Merck
All Photos(1)

Key Documents

EHU072911

Sigma-Aldrich

MISSION® esiRNA

MISSION® esiRNA

targeting human FOSL2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCTGGGGATTCAGCTCTAGAGGTCACAGTATCCTCGTTTGAAAGATAATTAAGATCCCCCGTGGAGAAAGCAGTGACACATTCACACAGCTGTTCCCTCGCATGTTATTTCATGAACATGACCTGTTTTCGTGCACTAGACACACAGAGTGGAACAGCCGTATGCTTAAAGTACATGGGCCAGTGGGACTGGAAGTGACCTGTACAAGTGATGCAGAAAGGAGGGTTTCAAAGAAAAAGGATTTTGTTTAAAATACTTTAAAAATGTTATTTCCTGCATCCCTTGGCTGTGATGCCCCTCTCCCGATTTCCCAGGGGCTCTGGGAGGGACCCTTCTAAGAAGATTGGGCAGTTGGGTTTCTGGCTTGAGATGAATCCAAGCAGCAGAATGAGCCAGGAGTAGCAGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Ling et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 104, 411-419 (2018-05-23)
Hepatocyte proliferation and apoptosis are critical cellular behaviors in rat liver as a result of a liver injury. Herein, we performed this study in order to evaluate the role of miR-30e and its target Fos-Related Antigen-2 (FOSL2) in septic rats
Shuo Li et al.
Experimental cell research, 373(1-2), 57-61 (2018-08-17)
Among different cancers, incidence and mortality of colorectal cancer (CRC) is one of the highest. KRAS mutation is one of the underlying features in the pathogenesis of CRC with CRC tumors harboring mutant KRAS exhibiting a more aggressive behavior compared
Poonam Sarode et al.
Science advances, 6(23), eaaz6105-eaaz6105 (2020-06-18)
Tumor-associated macrophages (TAMs) influence lung tumor development by inducing immunosuppression. Transcriptome analysis of TAMs isolated from human lung tumor tissues revealed an up-regulation of the Wnt/β-catenin pathway. These findings were reproduced in a newly developed in vitro "trained" TAM model.
Keito Okazaki et al.
Nature communications, 11(1), 5911-5911 (2020-11-22)
Transcriptional dysregulation, which can be caused by genetic and epigenetic alterations, is a fundamental feature of many cancers. A key cytoprotective transcriptional activator, NRF2, is often aberrantly activated in non-small cell lung cancers (NSCLCs) and supports both aggressive tumorigenesis and
Jon M Carthy et al.
Journal of cellular physiology, 230(12), 3084-3092 (2015-06-23)
Transforming growth factor-β (TGF-β) is a multifunctional cytokine which stimulates the differentiation of fibroblasts into myofibroblasts. Myofibroblasts are critical for normal wound healing, but also accumulate pathologically in a number of chronic inflammatory conditions where they are key contributors to

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service