Merck
  • Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Nature communications (2020-11-22)
Keito Okazaki, Hayato Anzawa, Zun Liu, Nao Ota, Hiroshi Kitamura, Yoshiaki Onodera, Md Morshedul Alam, Daisuke Matsumaru, Takuma Suzuki, Fumiki Katsuoka, Shu Tadaka, Ikuko Motoike, Mika Watanabe, Kazuki Hayasaka, Akira Sakurada, Yoshinori Okada, Masayuki Yamamoto, Takashi Suzuki, Kengo Kinoshita, Hiroki Sekine, Hozumi Motohashi
ABSTRACT
Transcriptional dysregulation, which can be caused by genetic and epigenetic alterations, is a fundamental feature of many cancers. A key cytoprotective transcriptional activator, NRF2, is often aberrantly activated in non-small cell lung cancers (NSCLCs) and supports both aggressive tumorigenesis and therapeutic resistance. Herein, we find that persistently activated NRF2 in NSCLCs generates enhancers at gene loci that are not normally regulated by transiently activated NRF2 under physiological conditions. Elevated accumulation of CEBPB in NRF2-activated NSCLCs is found to be one of the prerequisites for establishment of the unique NRF2-dependent enhancers, among which the NOTCH3 enhancer is shown to be critical for promotion of tumor-initiating activity. Enhancer remodeling mediated by NRF2-CEBPB cooperativity promotes tumor-initiating activity and drives malignancy of NRF2-activated NSCLCs via establishment of the NRF2-NOTCH3 regulatory axis.
Sigma-Aldrich
MISSION® esiRNA, targeting human MZF1
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human GABPA
Sigma-Aldrich
MISSION® esiRNA, targeting human FOSL2
Sigma-Aldrich
MISSION® esiRNA, targeting human NOTCH3
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid