Skip to Content
Merck
All Photos(1)

Key Documents

EHU052901

Sigma-Aldrich

MISSION® esiRNA

targeting human GDF15

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAGGTGCAAGTGACCATGTGCATCGGCGCGTGCCCGAGCCAGTTCCGGGCGGCAAACATGCACGCGCAGATCAAGACGAGCCTGCACCGCCTGAAGCCCGACACGGTGCCAGCGCCCTGCTGCGTGCCCGCCAGCTACAATCCCATGGTGCTCATTCAAAAGACCGACACCGGGGTGTCGCTCCAGACCTATGATGACTTGTTAGCCAAAGACTGCCACTGCATATGAGCAGTCCTGGTCCTTCCACTGTGCACCTGCGCGGAGGACGCGACCTCAGTTGTCCTGCCCTGTGGAATGGGCTCAAGGTTCCTGAGACACCCGATTCCTGCCCAAACAGCTGTATTTATATAAGTCTGTTATTTATTATTAATTTATTGGGGTGACCTTCTTGGGGACTCGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Louca et al.
Cancers, 11(8) (2019-08-16)
Glioblastoma multiforme (GBM) is the most aggressive type of brain tumor due to its invasive phenotype. Ras suppressor 1 (RSU-1) is a cell-extracellular matrix adhesion protein and we recently found that it promotes cell invasion in aggressive cells and inhibits
Yanwei Lu et al.
Cell death & disease, 8(9), e3036-e3036 (2017-09-08)
CDP138, a CDK5 binding partner, regulates cell proliferation and migration. However, the mechanisms by which CDP138 functions in these processes remain unclear. In this study, we show that CDP138 is frequently overexpressed and that high levels of CDP138 are correlated
Yixin Zhang et al.
Oncotarget, 8(22), 36531-36544 (2017-04-08)
Ischemia reperfusion (I/R) injury which inevitably occurs during heart transplantation is the major factor leading to organ failure and graft rejection. In order to develop new therapies to prevent I/R injury, we used both a murine heart transplantation model with
Kathrin Ackermann et al.
Atherosclerosis, 281, 128-136 (2019-01-19)
Growth differentiation factor-15 (GDF-15)/macrophage inhibitory cytokine-1 (MIC-1/GDF15) is associated with cardiovascular disease, inflammation and development of atherosclerosis and is highly expressed in macrophages (MΦ) of atherosclerotic lesions. Thus, we were interested in investigating the influence of GDF-15 in lipid homeostasis
Liang Chen et al.
Biochemical and biophysical research communications, 526(2), 293-299 (2020-03-27)
Ferroptosis is an iron-dependent form of regulated cell death. GDF15 affects various properties of cancer cells, but the role of GDF15 in ferroptosis has not been reported. In the present study, we found that GDF15 knockdown led to decreased expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service