Skip to Content
Merck
All Photos(1)

Key Documents

EHU109591

Sigma-Aldrich

MISSION® esiRNA

targeting human PHB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCTGAGCAACAGAAAAAGGCGGCCATCATCTCTGCTGAGGGCGACTCCAAGGCAGCTGAGCTGATTGCCAACTCACTGGCCACTGCAGGGGATGGCCTGATCGAGCTGCGCAAGCTGGAAGCTGCAGAGGACATCGCGTACCAGCTCTCACGCTCTCGGAACATCACCTACCTGCCAGCGGGGCAGTCCGTGCTCCTCCAGCTGCCCCAGTGAGGGCCCACCCTGCCTGCACCTCCGCGGGCTGACTGGGCCACAGCCCCGATGATTCTTAACACAGCCTTCCTTCTGCTCCCACCCCAGAAATCACTGTGAAATTTCATGATTGGCTTAAAGTGAAGGAAATAAAGGTAAAATCACTTCAGATCTCTAATTAGTCTATCAAATGAAACTCTTTCATTCTTCTCACATCCATCTACTTTTTTATCCACCTCCCTACCAAAAATTGCCAAGTGCCTATGCAAACCAGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijun Sun et al.
Biochemical and biophysical research communications, 513(2), 446-451 (2019-04-11)
Influenza virus infection is associated with type 1 diabetes (T1DM), but its pathogenesis remains unclear. Here, our study found that one of the monoclonal antibodies against H1N1 influenza virus hemagglutinin(HA) cross-reacted with human pancreatic tissue and further demonstrated that it
Debabrata Chowdhury et al.
Molecular and cellular biochemistry, 425(1-2), 155-168 (2016-11-18)
Numerous hypertrophic stimuli, including β-adrenergic agonists such as isoproterenol (ISO), result in generation of reactive oxygen species (ROS) and alteration in the mitochondrial membrane potential (Δψ) leading to oxidative stress. This process is well associated with phosphorylation of thymoma viral
Hajime Yurugi et al.
Journal of cell science, 133(12) (2020-06-06)
The RAS oncogenes are frequently mutated in human cancers and among the three isoforms (KRAS, HRAS and NRAS), KRAS is the most frequently mutated oncogene. Here, we demonstrate that a subset of flavaglines, a class of natural anti-tumour drugs and
Kinnosuke Yahiro et al.
Cellular microbiology, 21(8), e13033-e13033 (2019-04-23)
Vibrio cholerae produced-Cholix toxin (Cholix) is a cytotoxin that ADP-ribosylates eukaryotic elongation factor 2, inhibiting protein synthesis, and inducing apoptosis. Here, we identified prohibitin (PHB) 1 and 2 as novel Cholix-interacting membrane proteins in immortalised human hepatocytes and HepG2 cells
Satomi Miwa et al.
Nature communications, 5, 3837-3837 (2014-05-13)
Mitochondrial function is an important determinant of the ageing process; however, the mitochondrial properties that enable longevity are not well understood. Here we show that optimal assembly of mitochondrial complex I predicts longevity in mice. Using an unbiased high-coverage high-confidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service