Skip to Content
Merck
All Photos(1)

Key Documents

EHU002571

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC16A1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCCTTGAGAAAGCTGGAAAATCTGGTGTGAAAAAAGATCTGCATGATGCAAATACAGATCTTATTGGAAGACACCCTAAACAAGAGAAACGATCAGTCTTCCAAACAATTAATCAGTTCCTGGACTTAACCCTATTCACCCACAGAGGCTTTTTGCTATACCTCTCTGGAAATGTGATCATGTTTTTTGGACTCTTTGCACCTTTGGTGTTTCTTAGTAGTTATGGGAAGAGTCAGCATTATTCTAGTGAGAAGTCTGCCTTCCTTCTTTCCATTCTGGCTTTTGTTGACATGGTAGCCCGACCATCTATGGGACTTGTAGCCAACACAAAGCCAATAAGACCTCGAATTCAGTATTTCTTTGCGGCTTCCGTTGTTGCAAATGGAGTGTGTCATATGCTAGCACCTTTATCCACTACCTATGTTGGATTCTGTGTCTATGCGGGATTCTTTGGATTTGCCTTCGGGTGGCTCAGCTCCGTATTGTTTGAAACATTGATGGACCTTGTTGGACCCCAGAGGTTCTCCAGCGCTGTGGGATTGGTGACCATTGTGGAATGCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J De Saedeleer et al.
Oncogene, 33(31), 4060-4068 (2013-10-30)
The glycolytic end-product lactate is a pleiotropic tumor growth-promoting factor. Its activities primarily depend on its uptake, a process facilitated by the lactate-proton symporter monocarboxylate transporter 1 (MCT1). Therefore, targeting the transporter or its chaperon protein CD147/basigin, itself involved in
Jae Hoon Jeong et al.
Scientific reports, 8(1), 6672-6672 (2018-04-29)
Release of fatty acids from lipid droplets upon activation of the sympathetic nervous system (SNS) is a key step in nonshivering thermogenesis in brown adipose tissue (BAT). However, intracellular lipolysis appears not to be critical for cold-induced thermogenesis. As activation
Chunxiao Yan et al.
International journal of clinical and experimental pathology, 8(3), 2710-2718 (2015-06-06)
This study was designed to investigate the role of MCT1 in the development of cisplatin-resistant ovarian cancer and its possible relationship with Fas. We found the expression of MCT1 was obviously increased both in cisplatin-resistant ovarian cancer tissue and A2780/CP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service