Skip to Content
Merck
All Photos(1)

Key Documents

EMU012051

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Tnf

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTCCGGGCAGGTCTACTTTGGAGTCATTGCTCTGTGAAGGGAATGGGTGTTCATCCATTCTCTACCCAGCCCCCACTCTGACCCCTTTACTCTGACCCCTTTATTGTCTACTCCTCAGAGCCCCCAGTCTGTGTCCTTCTAACTTAGAAAGGGGATTATGGCTCAGAGTCCAACTCTGTGCTCAGAGCTTTCAACAACTACTCAGAAACACAAGATGCTGGGACAGTGACCTGGACTGTGGGCCTCTCATGCACCACCATCAAGGACTCAAATGGGCTTTCCGAATTCACTGGAGCCTCGAATGTCCATTCCTGAGTTCTGCAAAGGGAGAGTGGTCAGGTTGCCTCTGTCTCAGAATGAGGCTGGATAAGATCTCAGGCCTTCCTACCTTCAGACCTTTCCAGACTCTTCCCTGAGGTGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Pileczki et al.
International journal of molecular sciences, 14(1), 411-420 (2012-12-25)
Tumor necrosis factor alpha (TNF-α) is a pro-inflammatory cytokine involved in the promotion and progression of cancer, including triple negative breast cancer cells. Thus, there is significant interest in understanding the molecular signaling pathways that connect TNF-α with the survival
Hank Cheng et al.
Journal of neuroinflammation, 13, 19-19 (2016-01-27)
The basis for air pollution-associated neurodegenerative changes in humans is being studied in rodent models. We and others find that the ultrafine particulate matter (PM) derived from vehicular exhaust can induce synaptic dysfunction and inflammatory responses in vivo and in
Li Peng et al.
The International journal of artificial organs, 38(10), 565-571 (2015-11-07)
Periprosthetic osteolysis, involving RANK/RANKL/osteoprotegerin (OPG) and TNF-α/NFκB signaling, contributes to bone resorption and inflammation. We constructed lentivirus vectors to inhibit TNF-α and enhance OPG expression and assessed their impacts on wear debris-induced inflammation and osteoclastogenesis in an osteoclast/osteoblast coculture system.
Wenwen Zheng et al.
Molecular pain, 7, 40-40 (2011-05-24)
HIV-associated sensory neuropathy (HIV-SN) is one of the most common forms of peripheral neuropathy, affecting about 30% of people with acquired immune deficiency syndrome (AIDS). The symptoms of HIV-SN are dominated by neuropathic pain. Glia activation in the spinal cord
Praveen Thumbikat et al.
PLoS pathogens, 5(5), e1000415-e1000415 (2009-05-05)
Urinary tract infections are the second most common infectious disease in humans and are predominantly caused by uropathogenic E. coli (UPEC). A majority of UPEC isolates express the type 1 pilus adhesin, FimH, and cell culture and murine studies demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service