Skip to Content
Merck
All Photos(2)

Key Documents

E7908

Sigma-Aldrich

p3XFLAG-CMV-14 Expression Vector

shuttle vector for transient or stable intracellular expression of C-terminal 3xFLAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

3X FLAG tagged

grade

for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

The p3XFLAG-CMV-14 Expression Vector is a 6.3 kb derivative of pCMV51 used to establish expression of transient or stable intracellular C-terminal 3XFLAG fusion proteins in mammalian cells. The vector encodes three adjacent FLAG® epitopes (Asp-Tyr-Lys-Xaa-Xaa-Asp) downstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody. Efficiency of replication and genomic integration is optimal when using an SV40 T antigen-expressing host.

The p3XFLAG-CMV-7-BAP Control Plasmid is a 6.2 kb derivative of pCMV5 used for transient intracellular expression of N-terminal 3XFLAG bacterial alkaline phosphatase fusion protein in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp-Tyr-Lys-Xaa-Xaa-Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody. The third FLAG epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein.

Vector Maps and Sequences

Components

  • p3XFLAG-CMV-14 Expression Vector 20 μg (E4901) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • p3XFLAG-CMV-7-BAP Control Plasmid 20 μg (C7472) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG®-fusion constructs

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

This product is covered by the following patents owned by Sigma-Aldrich Co. LLC: US6,379,903, US7,094,548, JP4405125,EP1220933, CA2386471 and AU774216.
3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
p3xFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tom R Webb et al.
Endocrinology, 150(2), 720-726 (2008-09-27)
Melanocortin 2 receptor (MC2R) is the receptor for the pituitary hormone ACTH. When activated, MC2R stimulates cAMP production and adrenal steroidogenesis. The functional expression of the receptor requires melanocortin 2 receptor accessory protein (MRAP), a single-transmembrane domain protein involved in
Bengt Phung et al.
PloS one, 6(8), e24064-e24064 (2011-09-03)
The development of melanocytes is regulated by the tyrosine kinase receptor c-KIT and the basic-helix-loop-helix-leucine zipper transcription factor Mitf. These essential melanocyte survival regulators are also well known oncogenic factors in malignant melanoma. Despite their importance, not much is known
Qi-En Wang et al.
Cancer research, 72(3), 666-675 (2011-12-17)
XPC protein is a critical DNA damage recognition factor in nucleotide excision repair for which genetic deficiency confers a predisposition to cancer. In this study, we show that XPC has a function that is independent of its canonical function in
Zhi Sheng et al.
Molecular and cellular biology, 25(21), 9419-9426 (2005-10-18)
Fibroblast growth factor 2 (FGF-2), which is highly expressed in developing tissues and malignant cells, regulates cell growth, differentiation, and migration. Five isoforms (18 to approximately 34 kDa) of FGF-2 are derived from alternative initiation codons of a single mRNA.
Yan Qun Chen et al.
The Journal of biological chemistry, 283(15), 10048-10057 (2008-02-02)
AGPAT6 is a member of the 1-acylglycerol-3-phosphate O-acyltransferase (AGPAT) family that appears to be important in triglyceride biosynthesis in several tissues, but the precise biochemical function of the enzyme is unknown. In the current study, we show that AGPAT6 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service