Skip to Content
Merck
All Photos(1)

Key Documents

E6902

Sigma-Aldrich

Elastin, soluble from human aorta

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

biological source

human aorta

Quality Level

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

impurities

salt, free

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C27H48N6O6/c1-9-17(6)23(32-24(36)19(13-15(2)3)30-18(7)34)26(38)29-14-21(35)31-22(16(4)5)27(39)33-12-10-11-20(33)25(37)28-8/h15-17,19-20,22-23H,9-14H2,1-8H3,(H,28,37)(H,29,38)(H,30,34)(H,31,35)(H,32,36)/t17-,19-,20-,22-,23-/m0/s1

InChI key

DPUYCSDGMSDKKV-MKBYFEBXSA-N

Gene Information

human ... ELN(2006)

Biochem/physiol Actions

Elastin, along with fibrillin, is responsible for the elastic properties of skin and arteries. Reported inhibitor of Clostridial collagenase.
Reported inhibitor of Clostridial collagenase

Preparation Note

Prepared from E6777 using the method of Partrige, et al.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E H Sage et al.
Advances in experimental medicine and biology, 79, 291-312 (1977-01-01)
The aortae of a number of vertebrates and invertebrates were tested for the presence of elastin to determine when this protein first appears phylogenetically. Using several criteria, including amino acid composition, presence of desmosines, and histological characteristics, we found that
Jinzhi Li et al.
BMC cancer, 20(1), 217-217 (2020-03-17)
Colorectal cancer (CRC) is the most common cancer and a leading cause of death worldwide. Extracellular matrix (ECM) proteins regulate tumor growth and development in CRC. Elastin (ELN) is a component of ECM proteins involved in the tumor microenvironment. However
Hui Li et al.
Journal of anatomy, 236(1), 171-179 (2019-08-31)
Distinguishing arteries from veins in vivo has a great significance in clinical practices and preclinical studies. Optical imaging methods such as two-photon microscopy can provide high-resolution morphological information of tissue and are therefore extremely suitable for imaging small blood vessels.
The chemistry of connective tissues. 2. Soluble proteins derived from partial hydrolysis of elastin.
S M PARTRIDGE et al.
The Biochemical journal, 61(1), 11-21 (1955-09-01)
Aleksandra Kuzan et al.
Folia histochemica et cytobiologica, 59(1), 8-21 (2021-02-10)
Extracellular matrix (ECM) proteins have been associated with atherosclerotic complications, such as plaque rupture, calcification and aneurysm. It is not clear what role different types of collagen play in the pathomechanism of atherosclerosis. The aim of the study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service