Skip to Content
Merck
All Photos(1)

Key Documents

T0706

Sigma-Aldrich

Tropoelastin human

recombinant, expressed in E. coli, 1 mg

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$631.00

$631.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 MG
$631.00

About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.75

$631.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

description

1 mg recombinant human Tropoelastin as lyophilized powder.

sterility

electron beam irradiated

concentration

1 mg

impurities

≤10 EU/mL Endotoxin

storage temp.

−20°C

General description

Tropoelastin is a 60-72 kDa alternatively spliced extracellular matrix protein.[1] It is a soluble precursor of elastin and is distributed in elastic tissues.[2][3] Tropoelastin consists of an N- terminal 26-amino-acid signal peptide and highly basic strongly conserved C-terminal domain, that is required for both incorporation into elastin and for cellular interactions. It Is also characterized with a hydrophobic domain and exon 26A, which is an unusual highly hydrophilic domain.[2][1]

Application

Preparation Procedure
Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.

1. Add 1 ml of sterile 0.25% glacial acetic acid solution to the tropoelastin serum vial containing 1 mg.
2. Mix contents gently until material is completely solubilized. The solution may remain slightly hazy.
3. Transfer desired volume of solution from the serum vial to a dilution vessel if required. Further dilute to desired concentration using sterile 0.25% Acetic Acid solution. A typical working concentration may range from 1 to 50 μg/ml.
4. Add appropriate amount of diluted tropoelastin material to the culture surface.
5. Incubate at room temperature or 37 °C, covered, for approximately 1.5 hours.
6. After incubation, aspirate any remaining material.
7. Rinse coated surfaces carefully with sterile water and avoid scratching surfaces.
8. Coated surfaces are ready for use. They may also be stored at 2-10 °C damp or air dried if sterility is maintained.
Note: To maintain sterility, perform all operations in a laminar flow hood employing aseptic techniques.

Biochem/physiol Actions

Tropoelastin exhibits the property of coacervation and plays a vital role in fiber formation and assembly.[2][3] Abnormality in the tropoelastin is associated with the development of hereditary disease, cutis laxa (where the elasticity of the skin is lost).[2]

Features and Benefits

Tropoelastin available in its native state. Unique since it becomes cross-linked immediately after its synthesis by the cell and during its export into the extracellular matrix.

Quality

Identity and purity qualitatively determined by SDS-PAGE

Other Notes

Storage at -20 °C prior to solubilization is recommended. Storage at 2-10 °C after reconstitution is recommended.

Shelf life is 24 months when stored at -20 °C. Shelf life after reconstitution is 3 months when stored at 2-10 °C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemistry of tropoelastin.
Vrhovski B and Weiss A S
European Journal of Biochemistry, 258(1), 1-18 (1998)
Nasim Annabi et al.
Advanced materials (Deerfield Beach, Fla.), 28(1), 40-49 (2015-11-10)
A highly elastic hybrid hydrogel of methacryloyl-substituted recombinant human tropoelastin (MeTro) and graphene oxide (GO) nanoparticles are developed. The synergistic effect of these two materials significantly enhances both ultimate strain (250%), reversible rotation (9700°), and the fracture energy (38.8 ±
Domains in Tropoelastin That Mediate Elastin Deposition in Vitro and in Vivo.
Kozel, B., et al.
The Journal of Biological Chemistry, 2378 (20), 18491-18498 (2003)
Tropoelastin.
Wise S G and Weiss A S
The International Journal of Biochemistry & Cell Biology, 41(3), 494-497 (2009)
Coacervation of tropoelastin results in fiber formation.
Cox B A, et al.
The Journal of Biological Chemistry, 249(3), 997-998 (1974)

Questions

  1. Hi, Can you send me the sequence of this protein? do you have any peptide mapping or data confirming the sequence?

    1 answer
    1. The sequence of this product is not mapped.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service