Skip to Content
Merck
All Photos(1)

Key Documents

E6402

Sigma-Aldrich

Elastin from mouse lung

powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

Pricing and availability is not currently available. Contact Technical Service

biological source

mouse lung

Quality Level

form

powder

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C27H48N6O6/c1-9-17(6)23(32-24(36)19(13-15(2)3)30-18(7)34)26(38)29-14-21(35)31-22(16(4)5)27(39)33-12-10-11-20(33)25(37)28-8/h15-17,19-20,22-23H,9-14H2,1-8H3,(H,28,37)(H,29,38)(H,30,34)(H,31,35)(H,32,36)/t17-,19-,20-,22-,23-/m0/s1

InChI key

DPUYCSDGMSDKKV-MKBYFEBXSA-N

Gene Information

mouse ... Eln(13717)

General description

Elastin is a key protein of the extracellular matrix (ECM). It is made up of glycoproteins, glycosaminoglycans, collagen and proteoglycans. Elastin is mainly localized in the arteries and lungs.[1]
Elastin is an important extracellular matrix (ECM) protein derived from its precursor tropoelastin. Elastin is highly flexible than collagen.[2]

Application

Elastin mouse lung has been used as an extracellular matrix (ECM) protein for coating of plates.[3][4]

Biochem/physiol Actions

Elastin delivers elasticity to tissues and organs.[2][5] In addition, it is also involved in paracrine signaling. Genetic variation in the gene coding for elastin causes Williams-Beuren syndrome, cutis laxa and supravalvular aortic stenosis (SVAS).[5]
Elastin plays a critical role in alveolization in the fetal lung. In vivo radiochemical labelling indicates that mouse or rat lung elastin is synthesized primarily prenatally and during the first two weeks of life. It turns over very slowly, with a half-life estimated at several years.
Elastin plays an important role in the elasticity and resilience of many tissues, such as ligament, tendon and elastic cartilage. It is also involved in the elasticity of large arteries and skin.[1]

Preparation Note

Purified under mild extraction conditions to minimize peptide bond cleavage as described by Starcher, et al.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elastin
Biochemistry of Collagens, Laminins and Elastin, 290(2), 197-201 (2016)
Jiří Kanta
Frontiers in physiology, 7, 491-491 (2016-11-09)
A characteristic feature of liver cirrhosis is the accumulation of large amounts of connective tissue with the prevailing content of type I collagen. Elastin is a minor connective tissue component in normal liver but it is actively synthesized by hepatic
Elastin in the Liver
Kanta J.
Frontiers in Physiology, 7, 491-491 (2016)
An experimental system to study mechano transduction in fetal lung cells
Wang Y, et al.
Journal of Visualized Experiments, 7(60), e3543-e3543 (2012)
Yulian Wang et al.
Journal of visualized experiments : JoVE, (60) (2012-03-01)
Mechanical forces generated in utero by repetitive breathing-like movements and by fluid distension are critical for normal lung development. A key component of lung development is the differentiation of alveolar type II epithelial cells, the major source of pulmonary surfactant.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service