Skip to Content
Merck
All Photos(1)

Documents

E3020

Sigma-Aldrich

(±)-(E)-4-Ethyl-2-[(Z)-hydroxyimino]-5-nitro-3-hexen-1-yl-nicotinamide

≥97%

Synonym(s):

NOR-4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O4
CAS Number:
Molecular Weight:
306.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97%

storage temp.

−20°C

SMILES string

CC([N+]([O-])=O)/C(CC)=C/C(CNC(C1=CN=CC=C1)=O)=N/O

InChI

1S/C14H18N4O4/c1-3-11(10(2)18(21)22)7-13(17-20)9-16-14(19)12-5-4-6-15-8-12/h4-8,10,20H,3,9H2,1-2H3,(H,16,19)/b11-7+,17-13-

InChI key

KIWSYRHAAPLJFJ-DNZSEPECSA-N

Application

NO donor; releases nitric oxide spontaneously in a rate-controlled manner. Half-life in solution = 60 min.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Kita et al.
European journal of pharmacology, 257(1-2), 123-130 (1994-05-12)
(+-)-(E)-Ethyl-2-[(E)-hydroxyimino]-5-nitro-3-hexeneamide (FK409), which was isolated from microbial products, has been reported to show a vasorelaxant effect through a mechanism similar to that of the organic nitrates such as isosorbide dinitrate. In solution at pH 7.4, FK409 decomposed and released nitric

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service