Skip to Content
Merck
All Photos(2)

Key Documents

54918

Sigma-Aldrich

(S)-2-Hydroxybutyric acid

for chiral derivatization, ≥97.0%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$417.00

$417.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$417.00

About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1720940
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

$417.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

derivatization grade ((chiral))
for chiral derivatization

Assay

≥97.0% (T)
≥97.0%

form

solid

optical purity

enantiomeric ratio: ≥99:1 (GC)

mp

50-54 °C

storage temp.

2-8°C

SMILES string

CC[C@H](O)C(O)=O

InChI

1S/C4H8O3/c1-2-3(5)4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m0/s1

InChI key

AFENDNXGAFYKQO-VKHMYHEASA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Calderón et al.
Journal of chromatography. A, 1487, 194-200 (2017-02-01)
Chiral short chain aliphatic hydrocarboxylic acids (HCAs) are common compounds being part of different biological processes. In order to control and understand these processes is of pivotal importance to determine the identity of the involved enantiomer or their enantiomeric ratio.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service