Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
10 mg | Check Cart for Availability | $442.00 |
About This Item
Quality Segment
assay
≥98%
form
powder
solubility
DMSO: 10 mg/mL, clear, colorless to faintly yellow
storage temp.
−20°C
SMILES string
CC\C(=C/C(=N\O)C(N)=O)C(C)[N+]([O-])=O
InChI
1S/C8H13N3O4/c1-3-6(5(2)11(14)15)4-7(10-13)8(9)12/h4-5,13H,3H2,1-2H3,(H2,9,12)/b6-4+,10-7+
InChI key
MZAGXDHQGXUDDX-AGLLBGTNSA-N
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% | assay ≥97% | assay ≥98% (HPLC) | assay ≥98% (TLC) |
| form powder | form - | form powder | form powder |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. - | storage temp. - |
| solubility DMSO: 10 mg/mL, clear, colorless to faintly yellow | solubility - | solubility H2O: 15 mg/mL, clear | solubility 1 M HCl: 50 mg/mL, clear, colorless to faintly yellow |
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![(±)-(E)-4-Ethyl-2-[(E)-hydroxyimino]-5-nitro-3-hexenamide ≥98%](/deepweb/assets/sigmaaldrich/product/structures/404/859/0c6eb80d-2c3c-4bf1-98e8-1b319b597b12/640/0c6eb80d-2c3c-4bf1-98e8-1b319b597b12.png)
![(±)-(E)-4-Ethyl-2-[(Z)-hydroxyimino]-5-nitro-3-hexen-1-yl-nicotinamide ≥97%](/deepweb/assets/sigmaaldrich/product/structures/285/524/99543643-31d0-4c4f-b8da-9b1f872868ad/640/99543643-31d0-4c4f-b8da-9b1f872868ad.png)


