Skip to Content
Merck
All Photos(1)

Key Documents

E1407

Sigma-Aldrich

Monoclonal Anti-Erythropoietin antibody produced in rat

clone 148438, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-EP, Anti-EPO, Anti-Epo

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

148438, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

neutralization: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Epo(13856)

General description

Erythropoietin (Epo) is a pleotropic cytokine that promotes erythropoiesis or proliferation and differentiation of red blood cell precursors. Epo is mainly produced by liver cells in fetal stages, by kidneys under hypoxic conditions; liver and brain also secrete Epo is less amounts. The red blood cell production is stimulated when Epo binds and activates the Epo receptor (EpoR) dimer. EpoR directly regulates the signalling by Jak2, PI3K, NF-κB, STAT5A and STAT5B. Epo also inhibits apoptosis of erythroid progenitors by downregulating Fas and FasL.[1] The expression of Epo is under the control of the transcription factor hypoxia inducible factor (HIF). Erythropoietin exhibits disease-modifying effects in infectious diseases and chronic inflammatory disorders
Monoclonal Anti-Erythropoietin antibody specifically reacts with mouse Erythropoietin.

Immunogen

purified, CHO cell-derived recombinant mouse erythropoietin.

Application

Monoclonal Anti-Erythropoietin antibody is suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie S Watowich
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 59(7), 1067-1072 (2011-02-11)
The process of erythropoiesis in the fetal liver and adult bone marrow is regulated by the hormone erythropoietin (Epo), which is produced in the kidney at low levels under homeostatic conditions. Defects in Epo production result in severe anemia; use
Wolfgang Jelkmann
The Journal of physiology, 589(Pt 6), 1251-1258 (2010-11-17)
The hormone erythropoietin (Epo) maintains red blood cell mass by promoting the survival, proliferation and differentiation of erythrocytic progenitors. Circulating Epo originates mainly from fibroblasts in the renal cortex. Epo production is controlled at the transcriptional level. Hypoxia attenuates the
Johannes Vogel et al.
The Journal of physiology, 589(Pt 6), 1259-1264 (2011-02-02)
As the basic function of erythropoietin (Epo) is stimulation of red blood cell production, systemic overexpression of Epo results in erythrocytosis. The patho-physiological consequences of chronically elevated red blood cell counts have been studied in Epo overexpressing mice. Genetically modified
Manfred Nairz et al.
Microbes and infection, 14(3), 238-246 (2011-11-19)
Erythropoietin (EPO) is a multi-functional cytokine, which exerts erythropoietic effects but also carries anti-apoptotic and immune-modulatory activities upon binding to two distinct receptors which are expressed on erythroid, parenchymal and immune cells, respectively. Whereas EPO ameliorates hemolytic anemia in malaria
Manfred Nairz et al.
Immunity, 34(1), 61-74 (2011-01-25)
Erythropoietin (EPO) is the principal cytokine regulating erythropoiesis through its receptor, EPOR. Interestingly, EPORs are also found on immune cells with incompletely understood functions. Here, we show that EPO inhibits the induction of proinflammatory genes including tumor necrosis factor (TNF)-α

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service