Skip to Content
Merck
All Photos(1)

Key Documents

E0500

Sigma-Aldrich

Egg whites from chicken

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$338.00

$338.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$338.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

$338.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

chicken

form

powder

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... OVAL(396058)

Application

Egg whites from chicken have been used in studies of enzyme production of bioactive peptides that show strong antioxidant activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrine Bækby Graversen et al.
Frontiers in microbiology, 11, 496-496 (2020-04-16)
The intestinal gut microbiota is essential for maintaining host health. Concerns have been raised about the possible connection between antibiotic use, causing microbiota disturbances, and the increase in allergic and autoimmune diseases observed during the last decades. To elucidate the
Preparation of antioxidant peptide from egg white protein and improvement of its activities assisted by high-intensity pulsed electric field.
Lin S., et al.
Journal of the Science of Food and Agriculture (2011)
Purification and identification of antioxidant peptides from egg white protein hydrolysate.
Chen C., et al.
Amino Acids (2011)
Arielle Vallée Locke et al.
Scandinavian journal of immunology, 95(5), e13148-e13148 (2022-02-14)
The use of antibiotics as well as changes in the gut microbiota have been linked to development of food allergy in childhood. It remains unknown whether administration of a single clinically relevant antibiotic directly promotes food allergy development when administrated
Hoda M Eltaher et al.
Acta biomaterialia, 113, 339-349 (2020-06-20)
Combating necrosis, by supplying nutrients and removing waste, presents the major challenge for engineering large three-dimensional (3D) tissues. Previous elegant work used 3D printing with carbohydrate glass as a cytocompatible sacrificial template to create complex engineered tissues with vascular networks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service