Skip to Content
Merck
All Photos(6)

Key Documents

A5378

Sigma-Aldrich

Albumin from chicken egg white

lyophilized powder, ≥90% (agarose gel electrophoresis)

Synonym(s):

Albumin from hen egg white, Ovalbumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

chicken egg white

Quality Level

type

Grade III

Assay

≥90% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

44287 Da by calculation

purified by

chromatography

technique(s)

bioconjugation: suitable

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... OVAL(396058)

Looking for similar products? Visit Product Comparison Guide

General description

Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. From the amino acid sequence, the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.

Application

The product is useful as a marker (~45 kDa) for protein electrophoresis. It has been used to produce an animal model of asthma. Ovalbumin was used as medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophil. It was also used in a study testing intestinal macromolecule absorption in the fetal pig after infusion of colostrum in utero.
A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.

Biochem/physiol Actions

Differential scanning calorimetry indicates that chicken egg albumin denatures at 84 °C. Ovalbumin can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Tong et al.
Biotechnology progress, 24(3), 714-719 (2008-04-24)
Bioactive materials have been explored for a broad range of applications including biocatalysts, biosensors, antifouling membranes and other functional and smart materials. We report herein a unique method for preparation of bioactive materials through a spin coating process. Specifically, we
Xiaoxia Song et al.
iScience, 25(7), 104507-104507 (2022-06-21)
To realize the potential to use micro/nanorobots for targeted cancer therapy, it is important to improve their biocompatibility and targeting ability. Here, we report on drug-loaded magnetic microrobots capable of polarizing macrophages into the antitumor phenotype to target and inhibit
Qian He et al.
Frontiers in immunology, 11, 571248-571248 (2020-10-20)
As the entry sites of many pathogens such as human immunodeficiency virus (HIV), mucosal sites are defended by rapidly reacting resident memory T cells (TRM). TRMs represent a special subpopulation of memory T cells that persist long term in non-lymphoid
A D Nisbet et al.
European journal of biochemistry, 115(2), 335-345 (1981-04-01)
The complete amino acid sequence of hen ovalbumin, comprising 385 residues, has been determined. The sequence was deduced from the 17 cyanogen bromide fragments and from peptides derived by digestion with a number of proteolytic enzymes. The molecular weight of
Christianne Bandeira-Melo et al.
Journal of immunological methods, 276(1-2), 227-237 (2003-05-10)
Eosinophils contain several preformed cytokines within their specific granules. Therefore, without requiring them in de novo synthesis of cytokines, eosinophils can release quantities of granule-derived cytokines by highly regulated mechanisms. However, eosinophil "degranulation" is poorly understood, in part, because available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service