Skip to Content
Merck
All Photos(2)

Documents

DUO92020

Sigma-Aldrich

Duolink® In Situ PLA® Probe Anti-Human PLUS

Affinity purified Donkey anti-Human IgG (H+L)

Synonym(s):

in situ Proximity Ligation Assay, Protein Protein Interaction Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.41

biological source

donkey (Polyclonal)

antibody form

affinity purified immunoglobulin (Secondary antibody)

product line

Duolink®

species reactivity

human

technique(s)

immunofluorescence: suitable
proximity ligation assay: suitable

suitability

suitable for brightfield
suitable for fluorescence

shipped in

wet ice

storage temp.

2-8°C

Application

Duolink® proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.

This product can be applied to both the Duolink® In Situ Fluorescence Protocol and the Duolink® In Situ Brightfield Protocol depending on the detection reagents used.

Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting and more.

To perform a complete Duolink® PLA® in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. Other necessary reagents include a pair of PLA probes from different species (one PLUS and one MINUS), detection reagents, wash buffers, and mounting medium. Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.HRP is also available for brightfield detection.
Specificity

PLA probe anti-Human reacts with whole molecule human IgG and the light chains of other human immunoglobulin?s. The PLA Probe Anti-Human Plus has minimal cross-reactivity with bovine, chicken, goat, guinea pig, Syrian hamster, horse, mouse, rabbit, rat, and sheep serum proteins. A MINUS probe of a different species must be used simultaneously with this product. See our Product Selection Guide for more information.

Application Note

Two primary antibodies raised in different species are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC) or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® PLA in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list

Features and Benefits

  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Single molecule sensitivity due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Quicker and simpler than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Components

This product is comprised of the following:
  • 5x PLA Probe Anti-Human PLUS - Donkey anti-human secondary antibody conjugated to oligonucleotide PLUS
  • 1x Blocking Solution - Reagent for blocking of the sample
  • 1x Antibody Diluent - For dilution of PLA probes and primary antibodies

See datasheet for more information.

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Support information including tips and tricks, frequently asked questions, and basic troubleshooting.

Things to consider for preparation, setup and execution of the Duolink® assay protocol

Related Content

Applications to detect, quantify and visualize protein-protein interactions, post-translational modifications and low expression protein detection using proximity ligation assay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service