Skip to Content
Merck
All Photos(1)

Key Documents

DMPS5C

Sigma-Aldrich

Dimethylpolysiloxane

viscosity 500 cSt (25 °C)(lit.)

Synonym(s):

Polydimethylsiloxane

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$395.00
500 G
$433.00

$395.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$395.00
500 G
$433.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

$395.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

form

liquid

mol wt

~17,250

viscosity

500 cSt(25 °C)(lit.)

SMILES string

[Si](=O)(C)C

InChI

1S/C2H6OSi/c1-4(2)3/h1-2H3

InChI key

SEUDSDUUJXTXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
Dimethylpolysiloxane is used in protein chromatography and affininty chromatography. Dimethylpolysiloxane was used to determine that postprandial inflammatory response after ingestion of heated oils in obese persons is reduced by the presence of phenol compounds.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

609.8 °F - closed cup

Flash Point(C)

321 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mara I Orozco et al.
Journal of agricultural and food chemistry, 59(13), 7194-7202 (2011-06-08)
The influence of deep frying, mimicked by 20 heating cycles at 180 °C (each cycle from ambient temperature to 180 °C maintained for 5 min), on the unsaponifiable fraction of vegetable edible oils represented by three characteristic families of compounds
Vyacheslav N Fishman et al.
Chemosphere, 84(7), 913-922 (2011-07-09)
The applicability of three different Silphenylene Silicone co-polymer (Si-Arylene) GC stationary phases (J&W Scientific DB-5ms, Varian VF-5ms, and VF-Xms) has been evaluated for the separation of all 136 tetra- through the octa- chlorinated dibenzo-p-dioxins (PCDD) and chlorinated dibenzofurans (PCDF) from
A Perez-Herrera et al.
Molecular nutrition & food research, 56(3), 510-514 (2011-12-14)
Heating during the process of cooking alters the chemical properties of foods and may affect subsequent postprandial inflammation. We tested the effects of four meals rich in different oils subjected to heating on the postprandial inflammatory metabolism of peripheral blood

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service