Skip to Content
Merck
All Photos(1)

Key Documents

361879

Supelco

Dimethylpolysiloxane

set of analytical standards, molecular weight series

Synonym(s):

Polydimethylsiloxane

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$536.00

$536.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
$536.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$536.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

mol wt

Mw 100,000
Mw 270,000
Mw 40,000

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

SMILES string

[Si](=O)(C)C

InChI

1S/C2H6OSi/c1-4(2)3/h1-2H3

InChI key

SEUDSDUUJXTXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.[1]

Application

Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

609.8 °F - closed cup

Flash Point(C)

321 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robust and biodegradable elastomers based on corn starch and polydimethylsiloxane (PDMS).
Ceseracciu L
ACS Applied Materials & Interfaces, 7(6), 3742-3753 (2015)
Molecular weight distribution of poly (dimethylsiloxane) by combining matrix-assisted laser desorption/ionization time-of-flight mass spectrometry with gel-permeation chromatography fractionation.
Montaudo, Giorgio, et al.
Rapid Communications in Mass Spectrometry, 9.12, 1158-1163 (1995)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How much of each standard (mass or volume) is provided in this standard kit?

    1 answer
    1. Each standard included consists of 25 grams.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service