Skip to Content
Merck
All Photos(3)

Key Documents

D9655

Sigma-Aldrich

pBR322 Hae III Digest

for DNA electrophoresis

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$859.00

$859.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
0.1 MG
$859.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

$859.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

form

liquid

usage

1000 uses (0.5 mg size)
200 uses (0.1 mg size)

suitability

suitable for electrophoresis (DNA)

storage temp.

−20°C

General description

Sigma′s pBR322 Hae III Digest contains 22 fragments from 8 - 587 bp. Ten ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.

Application

Suitable for size determination of dsDNA by electrophoresis with either agarose or polyacrylamide gels.

Components

Sigma′s pBR322 Hae III Digest is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Other Notes

Contains 22 fragments, 8–587 bp
For optimal resolution, the recommended agarose gel concentration is 2.5.

Preparation Note

Plasmid pBR322 DNA was digested with Hae III to completion, then purified by phenol extraction.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Choose the appropriate markers and ladders for nucleic acid size determination of samples separated by electrophoresis. Determine size of DNA, RNA and PCR-generated fragments using agarose or polyacrylamide gels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service