D3812
DirectLoad™ 50 bp DNA Step Ladder
ready-to-use marker for DNA electrophoresis
Synonym(s):
DirectLoad™ 50 bp DNA Marker, Ready-to-use 50 bp DNA Marker, Ready-to-use DirectLoad™ 50 bp DNA Marker, Ready-to-use DirectLoad™ 50 bp DNA Step Ladder
Select a Size
$237.00
Select a Size
About This Item
$237.00
Recommended Products
Quality Level
form
(liquid)
usage
sufficient for 100 loads
suitability
suitable for electrophoresis (DNA)
storage temp.
−20°C
Related Categories
General description
Application
Features and Benefits
- Ready-to-load
- Easy-to-use
- Popular band sizes
Components
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.
Choose the appropriate markers and ladders for nucleic acid size determination of samples separated by electrophoresis. Determine size of DNA, RNA and PCR-generated fragments using agarose or polyacrylamide gels.
Protocols
Protocol using antibody mediated hot start polymerase. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.
JumpStart™ Taq DNA Polymerase is an antibody-inactivated, hot start enzyme.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service