Skip to Content
Merck
All Photos(1)

Key Documents

D9071

Sigma-Aldrich

Donitriptan monohydrochloride

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N5O2 · HCl
CAS Number:
Molecular Weight:
439.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.NCCc1c[nH]c2ccc(OCC(=O)N3CCN(CC3)c4ccc(cc4)C#N)cc12

InChI

1S/C23H25N5O2.ClH/c24-8-7-18-15-26-22-6-5-20(13-21(18)22)30-16-23(29)28-11-9-27(10-12-28)19-3-1-17(14-25)2-4-19;/h1-6,13,15,26H,7-12,16,24H2;1H

InChI key

ZXENQGQAPOYDOJ-UHFFFAOYSA-N

Biochem/physiol Actions

Donitriptan is a potent, selective 5-HT1B/1D agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hermann Mucke
Expert opinion on investigational drugs, 11(12), 1813-1820 (2002-11-30)
While oral formulations of agonists at the serotonin (5-HT) receptor subdivisions 5-HT(1B/1D)--the "triptan" class of drugs--dominate the market for migraine therapy and while still more triptans are under development, recent research efforts have also explored other pharmacological avenues, which are
V Limmroth et al.
Pain, 92(1-2), 101-106 (2001-04-27)
Calcitonin gene related peptide (CGRP) released from the C-fibers projecting from the trigeminal ganglion to the meninges has been suggested to play a crucial role in the pathophysiology of headache, particularly migraine. In humans it has been shown that CGRP
B Tom et al.
Cephalalgia : an international journal of headache, 22(1), 37-47 (2002-05-08)
We investigated the effects of donitriptan, which possesses a uniquely high affinity and efficacy at 5-HT1B/1D receptors, on carotid and systemic haemodynamics in anaes thetized pigs. Donitriptan (0.16-100 microg kg(-1), i.v.) dose-dependently decreased total carotid blood flow and vascular conductance
Rémon W M van den Broek et al.
European journal of pharmacology, 443(1-3), 125-132 (2002-06-05)
Donitriptan is a potent, high efficacy agonist at 5-HT(1B/1D) receptors. We investigated the contractile effects of donitriptan and sumatriptan on human isolated blood vessels of relevance to therapeutic efficacy in migraine (middle meningeal artery) and coronary adverse events (coronary artery).
Robert Letienne et al.
The Journal of pharmacology and experimental therapeutics, 305(2), 749-754 (2003-02-28)
The effects of donitriptan on systemic arterial-jugular venous oxygen saturation difference were evaluated in pentobarbitone-anesthetized pigs. Oxygen and carbon dioxide partial pressures in systemic arterial and jugular venous blood as well as hemoglobin oxygen saturation were determined by conventional blood

Articles

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service