Skip to Content
Merck
All Photos(1)

Key Documents

A8106

Sigma-Aldrich

Altanserin hydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,3-dihydro-2-thioxo-4(1H)-quinazolinone hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$195.00
50 MG
$760.00

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$195.00
50 MG
$760.00

About This Item

Empirical Formula (Hill Notation):
C22H22FN3O2S · HCl · xH2O
CAS Number:
Molecular Weight:
447.95 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >5 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

O.Cl.Fc1ccc(cc1)C(=O)C2CCN(CC2)CCN3C(=S)Nc4ccccc4C3=O

InChI

1S/C22H22FN3O2S.ClH.H2O/c23-17-7-5-15(6-8-17)20(27)16-9-11-25(12-10-16)13-14-26-21(28)18-3-1-2-4-19(18)24-22(26)29;;/h1-8,16H,9-14H2,(H,24,29);1H;1H2

InChI key

MFGBKMPKRJCLPK-UHFFFAOYSA-N

Application

Altanserin hydrochloride hydrate has been used in the pretreatment of rat brain samples and as a non-radioactive control in the calibration curve preparation for quantifying [18F]altanserin in high-performance liquid chromatography.[1]
Altanserin hydrochloride hydrate is a specific 5-HT2/serotonergic antagonist and has been used in a study to assess serotonin release capacity in the human brain.

Biochem/physiol Actions

Altanserin hydrochloride hydrate is a specific 5-HT2/serotonergic antagonist.
The radioligand of altanserin, [18F]altanserin is useful as a positron emission tomography (PET)-radioligand for the visualization and quantification of serotonin2A (5-HT2A) receptors.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirokazu Nagatani et al.
Journal of anesthesia, 25(2), 225-228 (2011-03-02)
Blockade of 5-hydroxytryptamine (5-HT)(2A) receptors reportedly mediates or modulates the ability of isoflurane to produce immobility during noxious stimulation and would thereby influence MAC (the minimum alveolar concentration required to suppress movement in response to noxious stimulation in 50% of
Patrick J Riss et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 31(12), 2334-2342 (2011-07-14)
The 5-hydroxytryptamine type 2a (5-HT(2A)) selective radiotracer [(18)F]altanserin has been subjected to a quantitative micro-positron emission tomography study in Lister Hooded rats. Metabolite-corrected plasma input modeling was compared with reference tissue modeling using the cerebellum as reference tissue. [(18)F]altanserin showed
Glenn Pauwelyn et al.
BMC veterinary research, 15(1), 415-415 (2019-11-23)
Currently, [18F] altanserin is the most frequently used PET-radioligand for serotonin2A (5-HT2A) receptor imaging in the human brain but has never been validated in dogs. In vivo imaging of this receptor in the canine brain could improve diagnosis and therapy
Hubertus Hautzel et al.
NeuroImage, 58(3), 905-911 (2011-07-05)
Behavioral and cellular studies indicate that serotonin interacting with the 5-HT2a receptor (5-HT2aR) is involved in cognitive processes supporting working memory (WM). However, 5-HT receptor neuroimaging studies directly relating WM-induced neuronal activations to concomitant changes in the availability of 5-HT
G Massarweh et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(11), 2040-2043 (2009-08-21)
[(18)F]Altanserin, an important PET radioligand for the in vivo imaging of the 5-HT(2A) receptor, was synthesized from its precursor nitro-altanserin in DMF or DMSO at high temperatures of 150 degrees C in an overall radiochemical yield (EOB) of 23-25% after

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service