Skip to Content
Merck

Skip To

D7283

3,5-Di-O-(p-toluyl)-2-deoxy-D-ribofuranosyl chloride

~90%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
$1,140.00

About This Item

Empirical Formula (Hill Notation):
C21H21ClO5
CAS Number:
Molecular Weight:
388.84
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
224-367-3
MDL number:

$1,140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~90%

Quality Segment

form

powder

mp

117-119  °C

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)C(=O)OC[C@H]2O[C@@H](Cl)C[C@@H]2OC(=O)c3ccc(C)cc3

InChI

1S/C21H21ClO5/c1-13-3-7-15(8-4-13)20(23)25-12-18-17(11-19(22)26-18)27-21(24)16-9-5-14(2)6-10-16/h3-10,17-19H,11-12H2,1-2H3/t17-,18+,19+/m0/s1

InChI key

FJHSYOMVMMNQJQ-IPMKNSEASA-N

Application

3,5-Di-O-(p-toluyl)-2-deoxy-D-ribofuranosyl is used as a precursor for the synthesis of unnatural β-D-ribofuranosyl containing nucleosides and nucleotides.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7087047439752926
assay

~90%

assay

90%

assay

≥90% (HPLC)

assay

≥90% (HPLC)

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

mp

117-119  °C

mp

238-244 °C

mp

-

mp

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters