Skip to Content
Merck
All Photos(2)

Key Documents

D6317

Sigma-Aldrich

DMeOB

≥98% (HPLC), solid

Synonym(s):

3-Methoxybenzaldehyde [(3-methoxyphenyl)methylene]hydrazone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$384.00

$384.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
50 MG
$384.00

About This Item

Empirical Formula (Hill Notation):
C16H16N2O2
CAS Number:
Molecular Weight:
268.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$384.00


Estimated to ship onMay 28, 2025


Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

DMSO: ~28 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

COc1cccc(\C=N\N=C\c2cccc(OC)c2)c1

InChI

1S/C16H16N2O2/c1-19-15-7-3-5-13(9-15)11-17-18-12-14-6-4-8-16(10-14)20-2/h3-12H,1-2H3/b17-11+,18-12+

InChI key

FBNPHFBYHYNMHC-JYFOCSDGSA-N

Biochem/physiol Actions

Negative allosteric modulator at the metabotropic glutamate receptor mGluR5.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie A O'Brien et al.
Molecular pharmacology, 64(3), 731-740 (2003-08-16)
We have identified a family of highly selective allosteric modulators of the group I metabotropic glutamate receptor subtype 5 (mGluR5). This family of closely related analogs exerts a spectrum of effects, ranging from positive to negative allosteric modulation, and includes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service