Skip to Content
Merck
All Photos(1)

Key Documents

D3758

Sigma-Aldrich

DL-2,4-Diaminobutyric acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2CH(NH2)COOH · 2HCl
CAS Number:
Molecular Weight:
191.06
Beilstein:
4708499
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

color

white

mp

197 °C (dec.) (lit.)

application(s)

detection

storage temp.

−20°C

SMILES string

Cl.Cl.NCCC(N)C(O)=O

InChI

1S/C4H10N2O2.2ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1H

InChI key

CKAAWCHIBBNLOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DL-2,4-Diaminobutyric acid is a racemic mixture of the selective GABA uptake inhibitors L-DABA and D-DABA. DL-2,4-diaminobutyric acid is found in cell-wall peptidoglycan of Leifsonia bigeumensis sp. Nov.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra M Leal et al.
Journal of neurobiology, 61(2), 189-208 (2004-09-25)
We have identified specific GABAergic-modulated behaviors in the juvenile stage of the fruit fly, Drosophila melanogaster via systemic treatment of second instar larvae with the potent GABA transport inhibitor DL-2,4-diaminobutyric acid (DABA). DABA significantly inhibited motor-controlled body wall and mouth
Amanda J Foss et al.
Toxicon : official journal of the International Society on Toxinology, 152, 150-159 (2018-08-14)
β-Methylamino-L-alanine (BMAA) has been identified as the potential cause of the amyotrophic lateral sclerosis/parkinsonism-dementia complex (ALS/PDC) observed in the Chamorro people of Guam. The principal hypothesis for BMAA exposure and intoxication relies on the biomagnification of BMAA in flying fox
Syed G Dastager et al.
International journal of systematic and evolutionary microbiology, 58(Pt 8), 1935-1938 (2008-08-05)
A Gram-positive, motile, mesophilic bacterium that formed yellowish-white, circular, convex, glistening colonies, strain MSL-27T, was isolated from a soil sample from Bigeum Island, Korea, and its taxonomic position was investigated by a polyphasic approach. Cells of strain MSL-27T are non-spore-forming
Aifeng Li et al.
Toxicon : official journal of the International Society on Toxinology, 151, 129-136 (2018-07-22)
In recent years, the neurotoxin β-N-methylamino-L-alanine (BMAA) has been reported in some marine mollusk species. To further discover BMAA in marine animals, a total of 59 samples belonging to 3 phyla, 22 families, and 43 species, were collected from Dalian
Shuqin Wang et al.
Toxicon : official journal of the International Society on Toxinology, 173, 39-47 (2019-11-18)
Neurotoxin β-N-methylamino-L-alanine (BMAA) has been widely detected in diverse aquatic organisms within different ecosystem trophic levels in recent years. It was the goal of this study to investigate the accumulation and tissue distribution of BMAA in marine opossum shrimp (Neomysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service