Skip to Content
Merck
All Photos(2)

Key Documents

150010

Sigma-Aldrich

Dimethyl 5-sulfoisophthalate sodium salt

98%

Synonym(s):

3,5-Bis(carbomethoxy)benzenesulfonic acid sodium salt, 3,5-Di(carbomethoxy)benzenesulfonic acid sodium salt, 5-Sulfoisophthalic acid dimethyl ester sodium salt, Dimethyl 5-(sodiosulfo)isophthalate, Dimethyl sulfoisophthalate sodium salt, Sodium 3,5-(dimethoxycarbonyl)benzenesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$105.00

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$105.00

About This Item

Linear Formula:
NaO3SC6H3-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
296.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

>300 °C (lit.)

functional group

ester
sulfonic acid

SMILES string

[Na+].COC(=O)c1cc(cc(c1)S([O-])(=O)=O)C(=O)OC

InChI

1S/C10H10O7S.Na/c1-16-9(11)6-3-7(10(12)17-2)5-8(4-6)18(13,14)15;/h3-5H,1-2H3,(H,13,14,15);/q;+1/p-1

InChI key

LLHSEQCZSNZLRI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl 5-sulfoisophthalate sodium salt was used to produce a series of water-soluble polymeric surfactants[1].

Application

  • Enhancing poly(lactic acid) crystallization: Dimethyl 5-sulfoisophthalate sodium salt was used as a nucleating agent in poly(lactic acid), significantly improving its crystallization behavior, which is crucial for developing biodegradable plastics with enhanced mechanical properties (Jongpanya-Ngam et al., 2022).
  • Development of biodegradable polyester ionomers: A study on biodegradable aliphatic polyester ionomers, potentially incorporating Dimethyl 5-sulfoisophthalate sodium salt, focused on synthesizing environmentally friendly materials that offer practical benefits in medical and packaging applications (Han et al., 2004).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and dispersion properties of water-soluble polyethylene glycol-dimethyl 5-sulfoisophthalate sodium salt polyester surfactants.
Lin LH, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 211(2), 173-178 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service