Skip to Content
Merck
All Photos(1)

Key Documents

D2947L

Sigma-Aldrich

Diffinity RapidTip®2

for PCR Purification with Polymerase Removal, Compatible with Rainin LTS® pipettes

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106300
NACRES:
NA.52

Quality Level

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Diffinity RapidTip2 effectively removes dNTPs, primers and primer dimers while providing greater than 90% recovery of pure DNA fragments from 100 bp to 10 kb. The functional pipette tip contains everything you need for PCR purification with polymerase removal. The tip is filled with a proprietary adsorption technology that has a differential affinity for PCR components. The impurities (polymerase, single strand primers, and nucleotides) are removed from the solution as it enters the tip. The RapidTip2 is optimized for a 50 μL PCR reaction sample, and the dispensed solution yields purified, high quality DNA ready for downstream applications such as DNA sequencing, SNP analysis, microarray printing, and T-A cloning.
Diffinity RapidTip2 effectively removes dNTPs, primers, primer dimers and DNA polymerase while providing greater than 90% recovery of pure DNA fragments from 100 bp to 10 kb. The functional pipette tip contains everything you need for PCR purification with polymerase removal. The tip is filled with a proprietary adsorption technology that has a differential affinity for PCR components. The dispensed solution yields purified, high quality DNA ready for downstream applications such as DNA sequencing, SNP analysis, microarray printing, and T-A cloning.

Features and Benefits

  • Single step
  • Recovers 90% of high quality dsDNA
  • Removes polymerase from PCR reactions and cloning
  • Optimized for 50 μL reaction

Legal Information

LTS is a registered trademark of Rainin Instrument, LLC
RapidTip is a registered trademark of Diffinity Genomics, Inc.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT RE 2 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kühlwein et al.
Journal of applied microbiology, 115(5), 1091-1106 (2013-07-31)
To assess the effects of dietary Saccharomyces cerevisiae β-(1,3)(1,6)-D-glucan supplementation (MacroGard(®)) on mirror carp (Cyprinus carpio L.) intestinal microbiota and ultrastructure of the enterocyte apical brush border. Carp were fed either a control diet or diets supplemented with 0.1, 1
Sarah L Dean et al.
Molecular ecology, 23(6), 1364-1378 (2013-10-12)
Nitrogen (N) deposition rates are increasing globally due to anthropogenic activities. Plant community responses to N are often attributed to altered competitive interactions between plants, but may also be a result of microbial responses to N, particularly root-associated fungi (RAF)
Teruaki Nakatsuji et al.
Nature communications, 4, 1431-1431 (2013-02-07)
Commensal microbes on the skin surface influence the behaviour of cells below the epidermis. We hypothesized that bacteria or their products exist below the surface epithelium and thus permit physical interaction between microbes and dermal cells. Here to test this
Michael J Goblirsch et al.
PloS one, 8(7), e69831-e69831 (2013-07-31)
A major hindrance to the study of honey bee pathogens or the effects of pesticides and nutritional deficiencies is the lack of controlled in vitro culture systems comprised of honey bee cells. Such systems are important to determine the impact
Myron T La Duc et al.
Applied and environmental microbiology, 78(16), 5912-5922 (2012-06-26)
Spacecraft hardware and assembly cleanroom surfaces (233 m(2) in total) were sampled, total genomic DNA was extracted, hypervariable regions of the 16S rRNA gene (bacteria and archaea) and ribosomal internal transcribed spacer (ITS) region (fungi) were subjected to 454 tag-encoded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service