Skip to Content
Merck
All Photos(1)

Documents

D1516

Sigma-Aldrich

Di-tert-butyl azodicarboxylate

Synonym(s):

Bis(1,1-dimethylethyl)azodicarboxylate, DBAD, Di-tert-butyl azodiformate, NSC 109889

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCON=NCOOC(CH3)3
CAS Number:
Molecular Weight:
230.26
Beilstein:
1911434
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

89-92 °C (lit.)

SMILES string

CC(C)(C)OC(=O)\N=N\C(=O)OC(C)(C)C

InChI

1S/C10H18N2O4/c1-9(2,3)15-7(13)11-12-8(14)16-10(4,5)6/h1-6H3/b12-11+

InChI key

QKSQWQOAUQFORH-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of hexapeptide key fragments via stereoselective selenocyclization/oxidative deselenylation or hydrazination/cyclization reactions
  • Asymmetric Michael addition reactions
  • Preparation of dipeptidyl peptidase IV dependent water-soluble prodrugs via Mitsunobu reaction
  • Synthesis of pyrroloisoquinoline template via stereoselective N-acyliminium-mediated cyclization and enolate amination for synthesis of peptidomimetic compounds
  • Barbier-type propargylation reactions
  • Synthesis of bacterial peptide deformylase (PDF) inhibitor fumimycin
  • Asymmetric amination of glycine Schiff bases

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service