Skip to Content
Merck
All Photos(1)

Key Documents

D9603

Sigma-Aldrich

Diallylamine

99%

Synonym(s):

Bis-allylamine, Bisallylamine, Di-2-propenylamine, N,N-Bis(allyl)amine, N-2-Propen-1-yl-2-propen-1-amine, N-2-propenyl-2-propen-1-amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$59.00
500 ML
$177.00
1 L
$220.00

$59.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$59.00
500 ML
$177.00
1 L
$220.00

About This Item

Linear Formula:
(CH2=CHCH2)2NH
CAS Number:
Molecular Weight:
97.16
Beilstein:
773718
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

3.35 (vs air)

vapor pressure

18 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.440 (lit.)

bp

111-112 °C (lit.)

mp

−88 °C (lit.)

density

0.787 g/mL at 25 °C (lit.)

SMILES string

C=CCNCC=C

InChI

1S/C6H11N/c1-3-5-7-6-4-2/h3-4,7H,1-2,5-6H2

InChI key

DYUWTXWIYMHBQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tawfik A Saleh et al.
Journal of colloid and interface science, 468, 324-333 (2016-02-09)
Hydrophobic cross-linked polyzwitterionic acid (HCPZA) containing long chain (C18) hydrophobes and residues of a glutamic acid having unquenched nitrogen valency was synthesized. Exploiting the chelating ability of the amino acid residues to scavenge toxic metals and the hydrophobic surface to
Yuanchen Liu et al.
Journal of hazardous materials, 368, 358-364 (2019-01-28)
We herein firstly presented supramolecularly imprinted polymeric (SMIP) solid phase microextraction (SPME) coatings which showed synergetic recognition for nitrophenols and bisphenol A. A series of β-cyclodextrins (β-CD) with different substituents were successfully designed and synthesized. It was employed as supramolecular
Yuanchen Liu et al.
Analytical and bioanalytical chemistry, 410(2), 509-519 (2017-12-01)
A series of β-cyclodextrin derivatives were designed and synthesized. The derivatives were investigated as functional monomers in molecularly imprinted polymeric solid-phase microextraction (MIP-SPME) fiber coatings. The coatings, with a layer thickness of 250 μm, were immobilized onto stainless steel using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service