Skip to Content
Merck
All Photos(1)

Key Documents

D0547

Sigma-Aldrich

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham

With L-glutamine and trace elements, without HEPES and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: no
sodium pyruvate: 0.11 g/L
HEPES: no
phenol red: yes
glucose: 3.15 g/L
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham is a 1:1 mixture.

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham has been used:

  • to culture cerebral cortices from mice embryos
  • as a component of the human corneal epithelium (HCE) media for culturing human limbal explants
  • to culture human corneal epithelial cell lines

Other Notes

Trace elements include ammonium molybdate, ammonium metavanadate, stannous chloride, sodium selenite, magnesium sulfate, nickel chloride, and sodium metasilicate

Quantity

Formulated to contain 12.0 grams of powder per liter of medium.

Reconstitution

Supplement with 1.2 g/L sodium bicarbonate

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimizing the role of limbal explant size and source in determining the outcomes of limbal transplantation: An in vitro study
Abhinav Reddy K, et al.
PLoS ONE (2017)
LPA2 promotes neuronal differentiation and neurite formation in neocortical development
Mizuki U, et al.
Biochemical and Biophysical Research Communications, 89-94 (2022)
Christo J Botha et al.
The Onderstepoort journal of veterinary research, 87(1), e1-e4 (2020-04-04)
Diplodiosis is an important neuromycotoxicosis of ruminants in South Africa when grazing on harvested maize fields in winter. It is believed to be caused by mycotoxin(s) synthesised by Stenocarpella (Diplodia) maydis. Although several metabolites have been isolated from S. maydis
Ceyda Acilan et al.
Biochimica et biophysica acta, 1861(2), 218-234 (2016-10-25)
To overcome the hurdles of cisplatin, majorly its toxicity and resistance, there has been extensive search for alternative anti-cancer metal-based compounds. Here, three Cu(II)-complexes, Cu(Sal-Gly)(phen), Cu(Sal-Gly)(pheamine), Cu(Sal-Gly)(phepoxy) are characterized for their interaction with DNA, cytotoxicity and mechanism of action. The
Selma M Soyal et al.
Molecular neurobiology, 57(2), 752-764 (2019-09-01)
PPARGC1A encodes a transcriptional co-activator also termed peroxisome proliferator-activated receptor (PPAR) gamma coactivator 1-alpha (PGC-1α) which orchestrates multiple transcriptional programs. We have recently identified CNS-specific transcripts that are initiated far upstream of the reference gene (RG) promoter. The regulation of

Articles

Nutrient Mixture F-12 Ham Formulation

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service