Skip to Content
Merck
All Photos(2)

Documents

D0162

Sigma-Aldrich

2′,3′-Dideoxyinosine

≥98% (HPLC)

Synonym(s):

ddI, ddIno

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O3
CAS Number:
Molecular Weight:
236.23
Beilstein:
3619529
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.55

Assay

≥98% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

OC[C@@H]1CC[C@@H](O1)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H12N4O3/c15-3-6-1-2-7(17-6)14-5-13-8-9(14)11-4-12-10(8)16/h4-7,15H,1-3H2,(H,11,12,16)/t6-,7+/m0/s1

InChI key

BXZVVICBKDXVGW-NKWVEPMBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nucleotide antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Ojewole et al.
Planta medica, 78(4), 354-361 (2011-12-14)
The buccal mucosal route offers several advantages but the delivery of certain drugs can be limited by low membrane permeability. This study investigated the buccal permeability properties of didanosine (ddI) and assessed the potential of Aloe vera gel (AVgel) as
Alejandro Arenas-Pinto et al.
Antiviral therapy, 17(2), 275-282 (2012-02-02)
Genetic predisposition to dideoxynucleoside-induced mitochondrial dysfunction might be related to mitochondrial DNA (mtDNA) polymorphisms. Severe hyperlactataemia is probably the best model to assess such a predisposition. For this exploratory study in White European and Black African HIV-infected adults, hypervariable region
Yuan-Zhen Xiong et al.
European journal of medicinal chemistry, 43(6), 1230-1236 (2007-09-18)
A series of novel 6-naphthyloxy substituted DATA analogues bearing different substituents on the C-6 position of triazine ring were synthesized and evaluated for their in vitro anti-HIV activity in MT-4 cells. The results demonstrated that most of the compounds in
Jan Balzarini et al.
European journal of medicinal chemistry, 42(7), 993-1003 (2007-02-27)
A series of novel thiazolidin-4-ones bearing a lipophilic adamantyl substituent at position 2, and versatile substituents on the nitrogen atom of the thiazolidine ring, were synthesized whereas several compounds exhibited a modest anti-HIV-1 activity, (+/-)-2-adamantan-1-yl-3-(4,6-dimethyl-pyridin-2-yl)-thiazolidin-4-one 22 was endowed with a
Jan Balzarini et al.
European journal of medicinal chemistry, 44(1), 303-311 (2008-04-19)
A series of novel thiazolidin-4-ones bearing a lipophilic adamantyl substituent at position 2 or 3 were synthesized. A majority of them showed a modest anti-HIV-1 activity, whereas 2-adamantan-1-yl-3-(4,6-dimethylpyrimidin-2-yl)-thiazolidin-4-one (8) was endowed with a remarkable antiviral potency (EC(50)=0.67 microM). The new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service